Merck
  • Amelioration of palmitate-induced metabolic dysfunction in L6 muscle cells expressing low levels of receptor-interacting protein 140.
Amelioration of palmitate-induced metabolic dysfunction in L6 muscle cells expressing low levels of receptor-interacting protein 140.
Canadian journal of physiology and pharmacology (2015-09-26)
Silvana Constantinescu, Lorraine P Turcotte
ABSTRACT
We have shown that reduced expression of receptor-interacting protein 140 (RIP140) alters the regulation of fatty-acid (FA) oxidation in muscle. To determine whether a high level of FA availability alters the effects of RIP140 on metabolic regulation, L6 myotubes were transfected with or without RNA interference oligonucleotide sequences to reduce RIP140 expression, and then incubated with high levels of palmitic acid, with or without insulin. High levels of palmitate reduced basal (53%-58%) and insulin-treated (24%-44%) FA uptake and oxidation, and increased basal glucose uptake (88%). In cells incubated with high levels of palmitate, low RIP140 increased basal FA uptake and insulin-treated FA oxidation and glucose uptake, and decreased basal glucose uptake and insulin-treated FA uptake. Under basal conditions, low RIP140 increased the mRNA content of FAT/CD36 (159%) and COX4 (61%), as well as the protein content of Nur77 (68%), whereas the mRNA expression of FGF21 (50%) was decreased, as was the protein content of CPT1b (35%) and FGF21 (44%). Under insulin-treated conditions, low RIP140 expression increased the mRNA content of MCAD (84%) and Nur77 (84%), as well as the protein content of Nur77 (23%). Thus, a low level of RIP140 restores the rates of FA uptake in the basal state, in part via a reduction in upstream insulin signaling. Our data also indicate that the protein expression of Nur77 may be modulated by RIP140 when muscle cells are metabolically challenged by high levels of palmitate.
Sigma-Aldrich
Ethidium bromide solution, for fluorescence, ~1% in H2O
Supelco
Pyridaben, PESTANAL®, analytical standard
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 500 μg/mL in H2O
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 10 mg/mL in H2O
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
SAFC
HEPES
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
SAFC
HEPES
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Potassium phosphate dibasic, anhydrous, for luminescence, for molecular biology, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, for molecular biology
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium phosphate dibasic, meets USP testing specifications
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Potassium phosphate dibasic, reagent grade, ≥98.0%
Sigma-Aldrich
Ethidium bromide, BioReagent, for molecular biology, powder
Sigma-Aldrich
Ethidium bromide, ~95% (HPLC)
Sigma-Aldrich
Magnesium sulfate solution, for molecular biology, 1.00 M±0.04 M
Sigma-Aldrich
L-Carnitine hydrochloride, synthetic, ≥98%
Sigma-Aldrich
Potassium phosphate dibasic solution, 1.0 M
Sigma-Aldrich
Potassium phosphate dibasic, ACS reagent, ≥98%
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥97%
Sigma-Aldrich
Potassium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Magnesium sulfate, anhydrous, reagent grade, ≥97%
Sigma-Aldrich
Potassium phosphate dibasic, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Magnesium sulfate, puriss. p.a., drying agent, anhydrous, ≥98.0% (KT), powder (very fine)