Merck
  • Substance P stimulates proliferation of spinal neural stem cells in spinal cord injury via the mitogen-activated protein kinase signaling pathway.
Substance P stimulates proliferation of spinal neural stem cells in spinal cord injury via the mitogen-activated protein kinase signaling pathway.
The spine journal : official journal of the North American Spine Society (2015-04-30)
Kyoung-Tae Kim, Hye-Jeong Kim, Dae-Chul Cho, Jae-Sung Bae, Seung-Won Park
ABSTRACT
Substance P (SP) is a neuropeptide that can influence neural stem/progenitor cell (NSPC) proliferation and neurogenesis in the brain. However, we could not find any experimental study that investigates SP action in the spinal cord. The aims of our study were to investigate the potential of the neuropeptide SP in promoting the proliferation of spinal cord-derived NSPCs (SC-NSPCs) after spinal cord injury (SCI) and to clarify the roles of the mitogen-activated protein (MAP) kinase signaling pathway in the process. This is a randomized animal study. The SC-NSPCs were suspended in 100 μL of a neurobasal medium containing SP (binds neurokinin-1 receptor [NK1R]) or L-703,606 (NK1R antagonist) and cultured in a 96-well plate for 5 days. A cell proliferation assay was performed using a 3-(4,5-dimethylthiazol-2-yl)-5-(3-carboxymethoxyphenyl)-2-(4-sulfophenyl)-2H-tetrazolium assay. A cord clipping method was used for the SCI model. Substance P and the NK1R antagonist (L-703,606) were infused intrathecally in SCI and sham models. Neural stem/progenitor cell proliferation was evaluated with immunostaining for bromodeoxyuridine (BrdU) and the immature neural marker nestin. An immunoblotting method was used for evaluating the MAP kinase signaling protein that contains extracellular signal-regulated kinases (ERKs and p38) and β-actin as the control group. In vitro, SP (0.01-10 μmol/L) increased the proliferation of cultured SC-NSPCs, with a peak increase of 35±2% at the 0.1 μmol/L concentration. Substance P of 0.1 μmol/L continuously increased SC-NSPC proliferation from 6 hours to 5 days, whereas the proliferation decreased from 18% to 98% with L-703,606 (1-10 μM). Intrathecal infusion of SP (1 μmol/L) for 7 days significantly increased the number of proliferating NPSCs (cells positive for both BrdU and nestin) in the spinal cord (by 120±17%, p<.05) in adult rats, but infusion of L-703,606 (10 μmol/L) significantly decreased the post-SCI induction of NPSC proliferation in the spinal cord (by 87±4%). Also, SP stimulates proliferation of SC-NSPCs via the MAP kinase signaling pathway, especially the phosphorylated ERK and phosphorylated p38 proteins. The phosphorylated ERK and phosphorylated p38 protein levels increased with SP (0.1 μmol/L, p<.05). These data indicate that SP can promote proliferation of SC-NSPCs in SCI and normal conditions and have important roles in neuronal regeneration after SCI. Also, ERKs and p38 MAP kinases are important signaling proteins in this process.
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
SAFC
Sodium chloride solution, 5 M
Supelco
Meprobamate solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, for molecular biology
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)