Merck
  • Alteration by p11 of mGluR5 localization regulates depression-like behaviors.
Alteration by p11 of mGluR5 localization regulates depression-like behaviors.
Molecular psychiatry (2015-09-16)
K-W Lee, L Westin, J Kim, J C Chang, Y-S Oh, B Amreen, J Gresack, M Flajolet, D Kim, A Aperia, Y Kim, P Greengard
ABSTRACT
Mood disorders and antidepressant therapy involve alterations of monoaminergic and glutamatergic transmission. The protein S100A10 (p11) was identified as a regulator of serotonin receptors, and it has been implicated in the etiology of depression and in mediating the antidepressant actions of selective serotonin reuptake inhibitors. Here we report that p11 can also regulate depression-like behaviors via regulation of a glutamatergic receptor in mice. p11 directly binds to the cytoplasmic tail of metabotropic glutamate receptor 5 (mGluR5). p11 and mGluR5 mutually facilitate their accumulation at the plasma membrane, and p11 increases cell surface availability of the receptor. Whereas p11 overexpression potentiates mGluR5 agonist-induced calcium responses, overexpression of mGluR5 mutant, which does not interact with p11, diminishes the calcium responses in cultured cells. Knockout of mGluR5 or p11 specifically in glutamatergic neurons in mice causes depression-like behaviors. Conversely, knockout of mGluR5 or p11 in GABAergic neurons causes antidepressant-like behaviors. Inhibition of mGluR5 with an antagonist, 2-methyl-6-(phenylethynyl)pyridine (MPEP), induces antidepressant-like behaviors in a p11-dependent manner. Notably, the antidepressant-like action of MPEP is mediated by parvalbumin-positive GABAergic interneurons, resulting in a decrease of inhibitory neuronal firing with a resultant increase of excitatory neuronal firing. These results identify a molecular and cellular basis by which mGluR5 antagonism achieves its antidepressant-like activity.
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
SAFC
HEPES
SAFC
HEPES
Sigma-Aldrich
MISSION® esiRNA, targeting mouse S100a10
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Calcium, dendritic pieces, purified by distillation, 99.99% trace metals basis
Sigma-Aldrich
Calcium, granular, 99%
Sigma-Aldrich
Calcium, pieces, <1 cm, 99%