- A human short open reading frame (sORF)-encoded polypeptide that stimulates DNA end joining.
A human short open reading frame (sORF)-encoded polypeptide that stimulates DNA end joining.
The Journal of biological chemistry (2014-03-13)
Sarah A Slavoff, Jinho Heo, Bogdan A Budnik, Leslyn A Hanakahi, Alan Saghatelian
PMID24610814
ABSTRACT
The recent discovery of numerous human short open reading frame (sORF)-encoded polypeptides (SEPs) has raised important questions about the functional roles of these molecules in cells. Here, we show that a 69-amino acid SEP, MRI-2, physically interacts with the Ku heterodimer to stimulate DNA double-strand break ligation via nonhomologous end joining. The characterization of MRI-2 suggests that this SEP may participate in DNA repair and underscores the potential of SEPs to serve important biological functions in mammalian cells.