Merck
Anxiogenic-like effects induced by hemopressin in rats.
Pharmacology, biochemistry, and behavior (2014-12-03)
Manoela V Fogaça, Andreza B Sonego, Vanessa Rioli, Fabio C Gozzo, Camila S Dale, Emer S Ferro, Francisco S Guimarães
ABSTRACT
Hemopressin (PVNFKFLSH; HP) is an orally active peptide derived from rat hemoglobin α-chain that could act as an inverse agonist at cannabinoid type 1 receptors (CB1). Here, we aim to investigate possible behavioral effects of HP in male Wistar rats tested in the elevated plus maze (EPM), following HP intraperitoneal (i.p., 0.05 mg/kg), oral (P.O., 0.05 and 0.5 mg/kg) or intracerebroventricular (I.C.V., 3 and 10 nmol) administration. HP induced a decrease in EPM open arm exploration, indicating an anxiogenic-like effect. However, i.p. administration of HP (1 mg/kg) followed by mass spectrometry analysis of brain-peptide extracts suggested that the intact HP does not cross the blood brain barrier. I.C.V. administrated HP produced anxiogenic-like effects that were prevented by Transient Receptor Potential Vanilloid Type 1 (TRPV1) antagonists, 6-iodonordihydrocapsaicin (1 nmol) or SB366791 (1 nmol), but not by the CB1 receptor antagonist AM251 (0.1 and 1 nmol). Altogether, these data suggest that I.C.V. administrated HP induces anxiogenic-like effects by activating TRPV1 receptors. The similar anxiogenic effects observed after i.p. or P.O. administration could be due to HP fragment(s) crossing the blood brain barrier. The present results advance our knowledge about HP pharmacology and suggest concerns in future clinical studies.
Sigma-Aldrich
6-Iodonordihydrocapsaicin, ≥98% (HPLC), solid
Sigma-Aldrich
Flunixin meglumine, ≥98% (HPLC)
Flunixin meglumine, European Pharmacopoeia (EP) Reference Standard
Supelco
Flunixin Meglumine, Pharmaceutical Secondary Standard; Certified Reference Material
Lysine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
AM251, >98% (HPLC), solid
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Supelco
L-Lysine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Supelco
L-Lysine monohydrochloride, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%