Merck
  • MIDA as a simple and highly efficient ligand for palladium-catalyzed Hiyama cross-coupling of aryl halides.
MIDA as a simple and highly efficient ligand for palladium-catalyzed Hiyama cross-coupling of aryl halides.
Organic & biomolecular chemistry (2014-08-12)
Mengping Guo, Liang Qi, Qiaochu Zhang, Zhiyong Zhu, Wei Li, Xiaogang Li
ABSTRACT
N-Methyliminodiacetic acid (MIDA) as a simple, air stable and water-soluble ligand has been used in the palladium-catalyzed Hiyama cross-coupling reaction of trimethoxyphenylsilane with aryl halides. The yield of the corresponding Hiyama coupling products is high up to around 90% in water and isopropanol under an ambient atmosphere in the presence of KOH and NaF.
Sigma-Aldrich
Methyliminodiacetic acid, 99%