Merck
  • Cajal body proteins differentially affect the processing of box C/D scaRNPs.
Cajal body proteins differentially affect the processing of box C/D scaRNPs.
PloS one (2015-04-16)
Isioma I Enwerem, Guowei Wu, Yi Tao Yu, Michael D Hebert
ABSTRACT
Small nuclear ribonucleoproteins (snRNPs), which are required for pre-mRNA splicing, contain extensively modified snRNA. Small Cajal body-specific ribonucleoproteins (scaRNPs) mediate these modifications. It is unknown how the box C/D class of scaRNPs localizes to Cajal Bodies (CBs). The processing of box C/D scaRNA is also unclear. Here, we explore the processing of box C/D scaRNA 2 and 9 by coilin. We also broaden our investigation to include WRAP53 and SMN, which accumulate in CBs, play a role in RNP biogenesis and associate with coilin. These studies demonstrate that the processing of an ectopically expressed scaRNA2 is altered upon the reduction of coilin, WRAP53 or SMN, but the extent and direction of this change varies depending on the protein reduced. We also show that box C/D scaRNP activity is reduced in a cell line derived from coilin knockout mice. Collectively, the findings presented here further implicate coilin as being a direct participant in the formation of box C/D scaRNPs, and demonstrate that WRAP53 and SMN may also play a role, but the activity of these proteins is divergent to coilin.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Stmn1
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Smn1
Sigma-Aldrich
MISSION® esiRNA, targeting human SMN1, SMN2
Sigma-Aldrich
MISSION® esiRNA, targeting human COIL
Sigma-Aldrich
MISSION® esiRNA, targeting human SNRPN
Sigma-Aldrich
MISSION® esiRNA, targeting human STMN1
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Wdr79
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Snrpn