Merck
  • Fabrication of enrofloxacin imprinted organic-inorganic hybrid mesoporous sorbent from nanomagnetic polyhedral oligomeric silsesquioxanes for the selective extraction of fluoroquinolones in milk samples.
Fabrication of enrofloxacin imprinted organic-inorganic hybrid mesoporous sorbent from nanomagnetic polyhedral oligomeric silsesquioxanes for the selective extraction of fluoroquinolones in milk samples.
Journal of chromatography. A (2014-08-31)
Hai-Bo He, Chen Dong, Bin Li, Jun-Ping Dong, Tian-Yu Bo, Tian-Lin Wang, Qiong-Wei Yu, Yu-Qi Feng
ABSTRACT
This paper reports a nanomagnetic polyhedral oligomeric silsesquioxanes (POSS)-directing strategy toward construction of molecularly imprinted hybrid materials for antibiotic residues determination in milk samples. The imprinted polymeric layer was facilely obtained through the copolymerization of active vinyl groups present on the nanomagnetic POSS (Fe3O4@POSS) surface and functional monomer (methacrylic acid) binding with template (enrofloxacin). Herein, the octavinyl POSS acted as not only the building blocks for hybrid rigid architectures but also the cross-linker for the formation of effective recognition sites during the imprinting process. The molecularly imprinted Fe3O4@POSS nanoparticles (Fe3O4@MI-POSS) demonstrated much higher adsorption capacity and selectivity toward enrofloxacin molecules and its analogs than the non-imprinted Fe3O4@POSS (Fe3O4@NI-POSS) materials. The imprinted particles were applied as a selective sorbent in solid-phase extraction focusing upon sample pretreatment in complex matrices prior to chromatographic analysis. The three FQs (ofloxacin, enrofloxacin, danofloxacin) could be selectively extracted from the biological matrix, while the matrix interferences were effectively eliminated simultaneously under the optimum extraction conditions. A simple, rapid and sensitive method based on the Fe3O4@MI-POSS material combined with HPLC-UV detection was then established for the simultaneous determination of three FQs from milk samples. The average recoveries of the three FQs were in the range of 75.6-108.9%. The relative standard deviations of intra- and inter-day ranging from 2.91 to 8.87% and from 3.6 to 11.5%, respectively. The limits of detections (S/N=3) were between 1.76 and 12.42 ng mL(-1). It demonstrates the effectiveness of trace analysis in complicated biological matrices utilizing magnetic separation in combination with molecularly imprinted solid-phase extraction, the rich chemistry of POSS makes it possible to be an ideal platform for generating molecular imprinted hybrid materials is also exhibited.
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Iron(III) chloride, anhydrous, powder, ≥99.99% trace metals basis
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
2,2′-Azobis(2-methylpropionitrile), purum, ≥98.0% (GC)
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
2,2′-Azobis(2-methylpropionitrile), 98%
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Iron(III) chloride, sublimed grade, ≥99.9% trace metals basis
Supelco
Phosphate Standard for IC, TraceCERT®, 1000 mg/L phosphate in water (nominal concentration)
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer