Merck
  • MiR-143 and MiR-145 regulate IGF1R to suppress cell proliferation in colorectal cancer.
MiR-143 and MiR-145 regulate IGF1R to suppress cell proliferation in colorectal cancer.
PloS one (2014-12-05)
Jiaojiao Su, Hongwei Liang, Weiyan Yao, Nan Wang, Suyang Zhang, Xin Yan, Hui Feng, Wenjing Pang, Yanbo Wang, Xueliang Wang, Zhen Fu, Yanqing Liu, Chihao Zhao, Junfeng Zhang, Chen-Yu Zhang, Ke Zen, Xi Chen, Yalei Wang
ABSTRACT
Insulin-like growth factor 1 receptor (IGF1R) is a transmembrane receptor that is activated by insulin-like growth factor 1 (IGF-1) and by a related hormone called IGF-2. It belongs to the large class of tyrosine kinase receptors and plays an important role in colorectal cancer etiology and progression. In this study, we used bioinformatic analyses to search for miRNAs that potentially target IGF1R. We identified specific target sites for miR-143 and miR-145 (miR-143/145) in the 3'-untranslated region (3'-UTR) of the IGF1R gene. These miRNAs are members of a cluster of miRNAs that have been reported to exhibit tumor suppressor activity. Consistent with the bioinformatic analyses, we identified an inverse correlation between miR-143/145 levels and IGF1R protein levels in colorectal cancer tissues. By overexpressing miR-143/145 in Caco2, HT29 and SW480 colorectal cancer cells, we experimentally validated that miR-143/145 directly recognizes the 3'-UTR of the IGF1R transcript and regulates IGF1R expression. Furthermore, the biological consequences of the targeting of IGF1R by miR-143/145 were examined by cell proliferation assays in vitro. We demonstrated that the repression of IGF1R by miR-143/145 suppressed the proliferation of Caco2 cells. Taken together, our findings provide evidence for a role of the miR-143/145 cluster as a tumor suppressor in colorectal cancer through the inhibition of IGF1R translation.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O