Merck
  • Magnolia officinalis attenuates free fatty acid-induced lipogenesis via AMPK phosphorylation in hepatocytes.
Magnolia officinalis attenuates free fatty acid-induced lipogenesis via AMPK phosphorylation in hepatocytes.
Journal of ethnopharmacology (2014-09-28)
Min Suk Seo, Sung-Woon Hong, Sung Hum Yeon, Young-Mok Kim, Key An Um, Jung Hwan Kim, Hye Jung Kim, Ki Churl Chang, Sang Won Park
ABSTRACT
Magnolia officinalis (MO) is a traditional Chinese herbal medicine that has been used in clinical practice to treat liver disease. The aim of this study is to examine the effects of MO on the development of nonalcoholic fatty liver in hepatocytes. Human hepatoma-derived HepG2 cells and mouse normal FL83B hepatocytes were exposed to 0.5mM free fatty acids (FFAs; oleate:palmitate, 2:1) for 24h to simulate conditions of nonalcoholic fatty liver in vitro. The cells were treated with a standardized MO extract 1h prior to FFA exposure. MO pretreatment attenuated the increases in intracellular lipid accumulation and triglyceride content in FFA-exposed hepatocytes in a dose-dependent manner. MO pretreatment significantly inhibited both sterol regulatory element-binding protein (SREBP)-1c activation and increases in fatty acid translocase, fatty acid synthase, and stearoyl CoA desaturase-1 protein expression in FFA-exposed hepatocytes in a dose-dependent manner. MO pretreatment markedly induced adenosine monophosphate-activated protein kinase (AMPK) phosphorylation in hepatocytes. Compound C, an AMPK inhibitor, blocked the inhibitory effect of MO on the increases in intracellular lipid accumulation and triglyceride content induced by FFAs. In hepatocytes pretreated with compound C, MO failed to inhibit SREBP-1c activation and the increases in fatty acid translocase, fatty acid synthase, and stearoyl-CoA desaturase-1 protein expression induced by FFAs. Our results indicate that MO attenuates triglyceride biosynthesis and accumulation induced by FFAs in hepatocytes, suggesting its pharmacological potential for the prevention of nonalcoholic fatty liver disease. These effects may be mediated by the inhibition of SREBP-1c via AMPK phosphorylation.
Product Description
Honokiol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Honokiol, ≥98% (HPLC), powder
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Supelco
Honokiol, analytical standard
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Glutathione, European Pharmacopoeia (EP) Reference Standard