Merck
  • Aqueous leaf extract of Jatropha gossypiifolia L. (Euphorbiaceae) inhibits enzymatic and biological actions of Bothrops jararaca snake venom.
Aqueous leaf extract of Jatropha gossypiifolia L. (Euphorbiaceae) inhibits enzymatic and biological actions of Bothrops jararaca snake venom.
PloS one (2014-08-16)
Juliana Félix-Silva, Thiago Souza, Yamara A S Menezes, Bárbara Cabral, Rafael B G Câmara, Arnóbio A Silva-Junior, Hugo A O Rocha, Ivanise M M Rebecchi, Silvana M Zucolotto, Matheus F Fernandes-Pedrosa
ABSTRACT
Snakebites are a serious public health problem due their high morbi-mortality. The main available specific treatment is the antivenom serum therapy, which has some disadvantages, such as poor neutralization of local effects, risk of immunological reactions, high cost and difficult access in some regions. In this context, the search for alternative therapies is relevant. Therefore, the aim of this study was to evaluate the antiophidic properties of Jatropha gossypiifolia, a medicinal plant used in folk medicine to treat snakebites. The aqueous leaf extract of the plant was prepared by decoction and phytochemical analysis revealed the presence of sugars, alkaloids, flavonoids, tannins, terpenes and/or steroids and proteins. The extract was able to inhibit enzymatic and biologic activities induced by Bothrops jararaca snake venom in vitro and in vivo. The blood incoagulability was efficiently inhibited by the extract by oral route. The hemorrhagic and edematogenic local effects were also inhibited, the former by up to 56% and the latter by 100%, in animals treated with extract by oral and intraperitoneal routes, respectively. The inhibition of myotoxic action of B. jararaca reached almost 100%. According to enzymatic tests performed, it is possible to suggest that the antiophidic activity may be due an inhibitory action upon snake venom metalloproteinases (SVMPs) and/or serine proteinases (SVSPs), including fibrinogenolytic enzymes, clotting factors activators and thrombin like enzymes (SVTLEs), as well upon catalytically inactive phospholipases A2 (Lys49 PLA2). Anti-inflammatory activity, at least partially, could also be related to the inhibition of local effects. Additionally, protein precipitating and antioxidant activities may also be important features contributing to the activity presented. In conclusion, the results demonstrate the potential antiophidic activity of J. gossypiifolia extract, including its significant action upon local effects, suggesting that it may be used as a new source of bioactive molecules against bothropic venom.
Sigma-Aldrich
Estropipate, ≥98% (HPLC), powder
Sigma-Aldrich
Snake venom from Bothrops jararaca
Supelco
Hexadecyltrimethylammonium bromide, suitable for ion pair chromatography, LiChropur
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, for molecular biology, ≥99.0% (AT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, for molecular biology, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥96.0% (AT)
Supelco
Hexadecyltrimethylammonium bromide, analytical standard
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
USP
Cetrimonium bromide, United States Pharmacopeia (USP) Reference Standard
Supelco
Sodium Citrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, for molecular biology, 1 M in H2O