Merck
  • Open flow microperfusion: pharmacokinetics of human insulin and insulin detemir in the interstitial fluid of subcutaneous adipose tissue.
Open flow microperfusion: pharmacokinetics of human insulin and insulin detemir in the interstitial fluid of subcutaneous adipose tissue.
Diabetes, obesity & metabolism (2014-09-23)
C Höfferer, D Tutkur, C Fledelius, C L Brand, T J Alsted, J Damgaard, E Nishimura, C B Jeppesen, S I Mautner, T R Pieber, F Sinner
ABSTRACT
To compare the time profile of insulin detemir and human insulin concentrations in the interstitial fluid (ISF) of subcutaneous adipose tissue during constant i.v. infusion and to investigate the relationship between the pharmacokinetics of both insulin molecules in plasma and the ISF of subcutaneous adipose tissue. During a 6-h hyperinsulinaemic-euglycaemic clamp (plasma glucose level 8 mmol/l) human insulin (21 and 42 pmol/min/kg) or insulin detemir (209 and 417 pmol/min/kg) were infused i.v. in eight rats per dose level. Open flow microperfusion (OFM) was used to continuously assess interstitial insulin concentrations in subcutaneous adipose tissue. At the lower infusion rate, insulin detemir appeared significantly later in the ISF than in the plasma (p < 0.05) and also appeared later in the ISF relative to human insulin (p < 0.005). By using OFM we were able to monitor albumin-bound insulin detemir directly in the ISF of subcutaneous tissue and confirm its delayed transendothelial passage to a peripheral site of action.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Albumin from rat serum, lyophilized powder, essentially fatty acid free, essentially globulin free, ≥99% (agarose gel electrophoresis)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Hydriodic acid, distilled, 57 wt. %, 99.999% trace metals basis
Sigma-Aldrich
Hydriodic acid, 57 wt. % in H2O, distilled, stabilized, 99.95%
Sigma-Aldrich
Hydriodic acid, contains no stabilizer, distilled, 57 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Hydriodic acid, contains no stabilizer, ACS reagent, 55%
Sigma-Aldrich
Hydriodic acid, contains ≤1.5% hypophosphorous acid as stabilizer, ACS reagent, ≥47.0%