Merck
  • Lipid and lipoprotein measurements and the risk of ischemic vascular events: Framingham Study.
Lipid and lipoprotein measurements and the risk of ischemic vascular events: Framingham Study.
Neurology (2015-01-09)
Aleksandra Pikula, Alexa S Beiser, Jing Wang, Jayandra J Himali, Margaret Kelly-Hayes, Carlos S Kase, Qiong Yang, Sudha Seshadri, Philip A Wolf
ABSTRACT
To examine the relationship between plasma lipid measurements and incident ischemic vascular events (ischemic stroke [IS], and as a positive control, myocardial infarction [MI]) in a community cohort. In 6,276 stroke-free Framingham participants (aged 64 ± 10 years, 56% female), we related plasma lipid levels (total cholesterol [TC], high-density lipoprotein cholesterol [HDL-C], and TC/HDL-C ratio) measured at the original cohort 15th (1977-1979) and 20th examination cycles (1986-1990) and (TC, HDL-C, TC/HDL-C ratio, triglycerides [TG], and low-density lipoprotein cholesterol [LDL-C]) measured at the offspring fourth examination (1995-1998), to 10-year risk of incident IS and MI. Utilizing genome-wide genotyping in the same subjects, we used mendelian randomization methods to assess whether observed associations were incidental or causal. During a mean follow-up of 9 years, 301 participants experienced incident IS. In multivariable-adjusted analyses, HDL-C ≤40 mg/dL and TC/HDL ratio ≥5 were associated with increased risk of IS (hazard ratio [95% confidence interval]: 1.59 [1.23-2.05], p < 0.001 and 1.47 [1.15-1.87], p < 0.001), but not TC or LDL-C. In adjusted analysis, a strong association between TG and IS was diminished. In the MI-free sample (n = 5,875, aged 64 ± 10 years, 58% female; 403 MI events), all lipid markers were associated with MI risk. A genetic risk score comprising 47 known determinants of circulating HDL-C was not associated with IS. In a middle-aged to elderly community sample, we observed that low HDL-C and high TC/HDL-C ratio, but not LDL-C or TG were associated with risk of incident IS. We observed the usual associations between lipids and risk of MI. Our findings suggest an important, but less likely causal, role of HDL-C over other lipid biomarkers for optimal stroke risk stratification.
Product Description
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
SAFC
Cholesterol, Plant-Derived, SyntheChol®