Merck
  • Micro-nanostructured CuO/C spheres as high-performance anode materials for Na-ion batteries.
Micro-nanostructured CuO/C spheres as high-performance anode materials for Na-ion batteries.
Nanoscale (2015-01-15)
Yanying Lu, Ning Zhang, Qing Zhao, Jing Liang, Jun Chen
ABSTRACT
In this paper, we report on the synthesis of micro-nanostructured CuO/C spheres by aerosol spray pyrolysis and their application as high-performance anodes in sodium-ion batteries. Micro-nanostructured CuO/C spheres with different CuO contents were synthesized through aerosol spray pyrolysis by adjusting the ratio of reactants and heat-treated by an oxidation process. The as-prepared CuO/C spheres show uniformly spherical morphology, in which CuO nanoparticles (∼10 nm) are homogeneously embedded in the carbon matrix (denoted as 10-CuO/C). The electrochemical performance of 10-CuO/C with a carbon weight of 44% was evaluated as the anode material for Na-ion batteries. It can deliver a capacity of 402 mA h g(-1) after 600 cycles at a current density of 200 mA g(-1). Furthermore, a capacity of 304 mA h g(-1) was obtained at a high current density of 2000 mA g(-1). The superior electrochemical performance of the micro-nanostructured CuO/C spheres leads to the enhancement of the electronic conductivity of the nanocomposite and the accommodation of the volume variation of CuO/C during charge/discharge cycling.
Product Description
Supelco
1-Methyl-2-pyrrolidinone, analytical standard
Sigma-Aldrich
1-Methyl-2-pyrrolidinone, suitable for HPLC, ≥99%
Sigma-Aldrich
1-Methyl-2-pyrrolidinone, ReagentPlus®, 99%
Sigma-Aldrich
1-Methyl-2-pyrrolidinone, biotech. grade, ≥99.7%
Sigma-Aldrich
1-Methyl-2-pyrrolidinone, ACS reagent, ≥99.0%