Merck
  • Determination of plasma protein binding of positron emission tomography radioligands by high-performance frontal analysis.
Determination of plasma protein binding of positron emission tomography radioligands by high-performance frontal analysis.
Journal of pharmaceutical and biomedical analysis (2014-06-13)
Nahid Amini, Ryuji Nakao, Magnus Schou, Christer Halldin
ABSTRACT
Positron emission tomography (PET) is an imaging technique based on the use of radioligands labeled with short lived radionuclides, such as (11)C (t½=20.4min) and (18)F (t½=109.8min), which as a consequence often requires rapid plasma protein binding analysis methods. In addition, PET radioligands can suffer from non-specific binding to the membrane when ultrafiltraion, which is the most commonly used method for measuring protein binding in PET, is employed. In this study a high-performance frontal analysis (HPFA) method based on incorporation of a gel filtration column (discovery(®) BIO GFC 100, 50mm×4.6mm, 5μm, 100Å) into a radio-LC system with phosphate buffered saline (PBS, pH 7.4) at a flow rate of 3ml/min as mobile phase was developed and investigated for four PET radioligands. The minimum injection volume (MIV) of plasma, which is a crucial factor in HPFA, was determined to be 200μl (human), 500μl (monkey), 700μl (human) and 1000μl (monkey) for these four radioligands. The MIV values increased as a higher fraction of the radioligand was present in the protein-free form. The protein binding results obtained were in good agreement with ultrafiltration and the method did not suffer from non-specific binding. The short analysis time (<12min) allowed multiple protein binding measurements during time course of a human [(11)C]PBR28 PET study.
Product Description
Supelco
Phosphate Standard for IC, TraceCERT®, 1000 mg/L phosphate in water (nominal concentration)
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Sigma-Aldrich
Sodium chloride solution, 0.85%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Potassium chloride, for molecular biology, ≥99.0%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Potassium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Supelco
Potassium chloride solution, ~3 M KCl, saturated with silver chloride
Supelco
Potassium chloride solution, conductance standard A acc. to ISO 7888, 0.1 M KCl
Supelco
Potassium chloride solution, BioUltra, ~3 M in H2O
Supelco
Potassium chloride solution, conductance standard C acc. to ISO 7888, 0.001 M KCl
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Supelco
ISA (ionic strength adjustment solution: 1 M KCl), 1 M KCl
Sigma-Aldrich
Potassium chloride, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Potassium chloride, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium chloride, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Potassium chloride, puriss. p.a., ≥99.5% (AT)