Merck
  • Quantitative determination of microbicidal spermicide 'nonoxynol-9' in rabbit plasma and vaginal fluid using LC-ESI-MS/MS: application to pharmacokinetic study.
Quantitative determination of microbicidal spermicide 'nonoxynol-9' in rabbit plasma and vaginal fluid using LC-ESI-MS/MS: application to pharmacokinetic study.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-07-13)
Yashpal S Chhonker, Hardik Chandasana, Veenu Bala, Lokesh Kumar, Vishnu Lal Sharma, Gopal Gupta, Rabi S Bhatta
ABSTRACT
Nonoxynol-9 (N-9), a microbicidal spermicide, has been in use as an over-the-counter contraceptive since the 1960s. A detailed account of its pharmacokinetic profile using highly sensitive detection method has not been reported yet. We developed and validated a rapid, selective and sensitive high-performance liquid chromatography coupled with tandem mass spectrometry (LC-MS/MS) method for N-9 detection in plasma and simulated vaginal fluid. The analytes were quantified using reverse phase Thermo Accucore C18 (150 mm × 4.6mm, 5 μm) column with isocratic elution using acetonitrile: 0.1% formic acid in triple distilled water (90:10, v/v) as mobile phase. The ionization was optimized using ESI (+) and selectivity was achieved by tandem mass spectrometric analysis using MRM transition, m/z 617.4→133.2 for N-9 and m/z 180.1→138.1 for phenacetin. The method was linear over the range 0.195-100 ng/mL. The method was accurate and precise with intra-batch and inter-batch accuracy (% bias) of less than ± 15% and precision (% CV) of <15% for N-9. The mean peak plasma concentration (Cmax) 4.87 ± 0.37 ng/mL was achieved 1.0h after vaginal application with terminal half-life 1.45 ± 0.07 h in rabbits. The validated method was successfully applied for pharmacokinetic study of N-9 in rabbits after vaginal administration.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
USP
Phenacetin Melting Point Standard, United States Pharmacopeia (USP) Reference Standard
Supelco
Phenacetin melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phenacetin, ≥98.0% (HPLC)
Supelco
Phenacetin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Magnesium chloride hexahydrate, meets USP testing specifications
Sigma-Aldrich
Magnesium chloride hexahydrate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Magnesium chloride hexahydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Magnesium chloride hexahydrate, tested according to Ph. Eur.
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Magnesium chloride hexahydrate, BioUltra, for molecular biology, ≥99.0% (KT)
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Magnesium chloride hexahydrate, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Magnesium chloride hexahydrate, 99.995% trace metals basis
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)