Merck
  • Antioxidant capacity of arabinoxylan oligosaccharide fractions prepared from wheat aleurone using Trichoderma viride or Neocallimastix patriciarum xylanase.
Antioxidant capacity of arabinoxylan oligosaccharide fractions prepared from wheat aleurone using Trichoderma viride or Neocallimastix patriciarum xylanase.
Food chemistry (2014-08-26)
Lovemore Nkhata Malunga, Trust Beta
ABSTRACT
The effect of xylanase type (Trichoderma viride or Neocallimastix patriciarum) and graded ethanol fractionation on the antioxidant capacity (AOC) of arabinoxylan oligosaccharides (AXOS) obtained from wheat aleurone was investigated. AXOS yields were higher using N. patriciarum (62%) than T. viride (44%). The fraction (F100) collected at >80% ethanol concentration constituted 60% of total recovered AXOS. Degree of substitution ranged from 0.20 to 0.60 for ethanol graded fractions. Ferulic acid (FA) esterified to AXOS (8.0 μg/ mg) was 2-fold lower for the N. patriciarum treatment. The mean AOC (41.6, 183.1, and 394.9 μM TE/mg) of T. viride treated AXOS was >1.4-fold higher than N. patriciarum treatment using DPPH and ABTS and ORAC assays, respectively. Fraction F100 had highest AOC. AOC was influenced by the content of esterified FA (R(2)=0.94). The type of xylanase had a major influence on the AOC of the resultant AXOS rich in FA content.
Product Description
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-(+)-Xylose, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Supelco
trans-Ferulic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
D-(+)-Xylose, ≥99% (GC)
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
1,1-Diphenyl-2-picrylhydrazine, 97%
Sigma-Aldrich
trans-Ferulic acid, 99%
Supelco
trans-Ferulic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)
Sigma-Aldrich
D-(+)-Xylose, ≥99%
Ferulic acid, European Pharmacopoeia (EP) Reference Standard
Fluorescein, European Pharmacopoeia (EP) Reference Standard
USP
Dextrose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
2,2-Diphenyl-1-picrylhydrazyl
Sigma-Aldrich
Fluorescein, for fluorescence, free acid
Sigma-Aldrich
trans-Ferulic acid, ≥99%
Sigma-Aldrich
D-(+)-Xylose, BioXtra, ≥99% (GC)
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Galactose, ≥98% (HPLC)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Galactose, meets analytical specification of Ph. Eur., BP
Millipore
D-(+)-Galactose, suitable for microbiology, ≥99.0%
Millipore
D-(−)-Arabinose, suitable for microbiology, ≥99.0%
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
USP
Galactose, United States Pharmacopeia (USP) Reference Standard
Supelco
Galactose, Pharmaceutical Secondary Standard; Certified Reference Material
Galactose, European Pharmacopoeia (EP) Reference Standard