Merck
  • Genotoxicity of tri- and hexavalent chromium compounds in vivo and their modes of action on DNA damage in vitro.
Genotoxicity of tri- and hexavalent chromium compounds in vivo and their modes of action on DNA damage in vitro.
PloS one (2014-08-12)
Zhijia Fang, Min Zhao, Hong Zhen, Lifeng Chen, Ping Shi, Zhiwei Huang
ABSTRACT
Chromium occurs mostly in tri- and hexavalent states in the environment. Hexavalent chromium [Cr(VI)] compounds are extensively used in diverse industries, and trivalent chromium [Cr(III)] salts are used as micronutrients and dietary supplements. In the present work, we report that they both induce genetic mutations in yeast cells. They both also cause DNA damage in both yeast and Jurkat cells and the effect of Cr(III) is greater than that of Cr(VI). We further show that Cr(III) and Cr(VI) cause DNA damage through different mechanisms. Cr(VI) intercalates DNA and Cr(III) interferes base pair stacking. Based on our results, we conclude that Cr(III) can directly cause genotoxicity in vivo.
Sigma-Aldrich
Chromium, chips, 99.995% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chromium(VI) oxide, 99.99% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chromium, powder, ≥99% trace metals basis, <45 μm
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chromium, powder, 99.5%, −100 mesh
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chromium, chips, thickness ~2 mm, 99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chromium(III) chloride, anhydrous, 99.99% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chromium(III) chloride, purified by sublimation, 99%
SKU
Pack Size
Availability
Price
Quantity
Chromium, foil, not light tested, 50x50mm, thickness 0.025mm, permanent polyester support, 99.99+%
SKU
Pack Size
Availability
Price
Quantity
Chromium, foil, not light tested, 25x25mm, thickness 0.01mm, permanent polyester support, 99.99+%
SKU
Pack Size
Availability
Price
Quantity
Chromium, foil, 25x25mm, thickness 2.0mm, hard, 99.7+%
SKU
Pack Size
Availability
Price
Quantity
Chromium, foil, light tested, 25x25mm, thickness 0.025mm, permanent polyester support, 99.99+%
SKU
Pack Size
Availability
Price
Quantity
Chromium, foil, 50x50mm, thickness 2.0mm, hard, 99.7+%
SKU
Pack Size
Availability
Price
Quantity
Chromium, foil, light tested, 25x25mm, thickness 0.02mm, permanent polyester support, 99.99+%
SKU
Pack Size
Availability
Price
Quantity
Chromium, foil, 50x50mm, thickness 3.0mm, hard, 99.95%
SKU
Pack Size
Availability
Price
Quantity
Chromium, microfoil, disks, 10mm, thinness 0.5μm, specific density 355μg/cm2, permanent mylar 3.5μm support, 99.99+%
SKU
Pack Size
Availability
Price
Quantity
Chromium, foil, not light tested, 25x25mm, thickness 0.0125mm, permanent polyester support, 99.99+%
SKU
Pack Size
Availability
Price
Quantity
Chromium, rod, 50mm, diameter 5.0mm, 99.7+%
SKU
Pack Size
Availability
Price
Quantity
Chromium, rod, 10mm, diameter 5.0mm, 99.7+%
SKU
Pack Size
Availability
Price
Quantity
Chromium, foil, not light tested, 25x25mm, thickness 0.009mm, permanent polyester support, 99.99+%
SKU
Pack Size
Availability
Price
Quantity
Chromium, foil, light tested, 25x25mm, thickness 0.05mm, permanent polyester support, 99.99+%
SKU
Pack Size
Availability
Price
Quantity
Chromium, bar, 100mm x 2mm x 2mm, 99.7+%
SKU
Pack Size
Availability
Price
Quantity
Chromium, foil, not light tested, 25x25mm, thickness 0.006mm, permanent polyester support, 99.99+%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chromium(VI) oxide, ReagentPlus®, 99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Chromium(VI) oxide, ACS reagent, ≥98.0%
SKU
Pack Size
Availability
Price
Quantity
Chromium, foil, not light tested, 50x50mm, thickness 0.03mm, permanent polyester support, 99.99+%
SKU
Pack Size
Availability
Price
Quantity
Chromium, foil, not light tested, 25x25mm, thickness 0.03mm, permanent polyester support, 99.99+%
SKU
Pack Size
Availability
Price
Quantity
Chromium, microfoil, disks, 25mm, thinness 0.25μm, specific density 178.7μg/cm2, permanent mylar 3.5μm support, 99.99+%
SKU
Pack Size
Availability
Price
Quantity
Chromium, foil, not light tested, 50x50mm, thickness 0.02mm, permanent polyester support, 99.99+%
SKU
Pack Size
Availability
Price
Quantity
Chromium, microfoil, disks, 25mm, thinness 0.5μm, specific density 355μg/cm2, permanent mylar 3.5μm support, 99.99+%
SKU
Pack Size
Availability
Price
Quantity
Chromium, foil, not light tested, 25x25mm, thickness 0.015mm, permanent polyester support, 99.99+%