Merck
  • Peptide Lv augments L-type voltage-gated calcium channels through vascular endothelial growth factor receptor 2 (VEGFR2) signaling.
Peptide Lv augments L-type voltage-gated calcium channels through vascular endothelial growth factor receptor 2 (VEGFR2) signaling.
Biochimica et biophysica acta (2015-02-24)
Liheng Shi, Soyoung Ko, Michael L Ko, Andy Jeesu Kim, Gladys Y-P Ko
ABSTRACT
We previously identified peptide Lv, a novel bioactive peptide that enhances the activity of L-type voltage-gated calcium channels (L-VGCCs) in cone photoreceptors. In this study, we verified that peptide Lv was able to augment L-VGCC currents in cardiomyocytes, as well as promote proliferation of endothelial cells. We used a proteomics approach to determine the specific receptors and binding partners of peptide Lv and found that vascular endothelial growth factor receptor 2 (VEGFR2) interacted with peptide Lv. Peptide Lv treatment in embryonic cardiomyocytes stimulated tyrosine autophosphorylation of VEGFR2 and activated its downstream signaling. Peptide Lv activity was blocked by DMH4, a VEGFR2 specific blocker, but not by SCH202676, an allosteric inhibitor of G protein-coupled receptors, suggesting that the activity of peptide Lv was mediated through VEGFR2 signaling. Inhibition of VEGFR tyrosine kinase or its downstream signaling molecules abolished the augmentation of L-VGCCs elicited by peptide Lv in cardiomyocytes. In addition, peptide Lv promoted cell proliferation of cultured human endothelial cells. Calcium entry through L-VGCCs is essential for excitation-contraction coupling in cardiomyocytes. Since peptide Lv was able to augment L-VGCCs through activation of VEGF signaling in cardiomyocytes and promote proliferation of endothelial cells, peptide Lv may play an important role in regulating the cardiovascular system.
Product Description
Millipore
Bifido Selective Supplement B, suitable for microbiology
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
Chelerythrine chloride, ≥95% (TLC), powder
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Retinol, ≥95.0% (HPLC), ~2700 U/mg
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Retinol, BioXtra, ≥97.5% (HPLC), ~3100 U/mg
Sigma-Aldrich
Retinol, synthetic, ≥95% (HPLC), (Powder or Powder with Lumps)
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC