Merck
  • Involvement of chromosome region maintenance 1 (CRM1) in the formation and progression of esophageal squamous cell carcinoma.
Involvement of chromosome region maintenance 1 (CRM1) in the formation and progression of esophageal squamous cell carcinoma.
Medical oncology (Northwood, London, England) (2014-08-26)
Xiaojing Yang, Lei Cheng, Li Yao, Hanru Ren, Shu Zhang, Xiao Min, Xueyu Chen, Jianguo Zhang, Mei Li
ABSTRACT
Chromosome region maintenance 1 (CRM1) has been related to several malignancies. The predictive value of CRM1 in the malignance and prognosis of esophageal squamous cell carcinoma (ESCC), however, is not clear yet. In this study, we displayed that CRM1 expression was up-regulated in ESCC using immunohistochemistry and Western blot. Statistical analysis demonstrated that patients with high CRM1 levels indicated shorter survival period. We further found that silencing CRM1 caused apoptosis in ESCC cell lines. Moreover, knockdown of CRM1 disturbed the expression of tumor suppressor proteins and inhibited NF-κB activity in ESCC cell lines, especially if the cell line was treated with 5-fluorouracil. In consequence, our results for the first time indicated that CRM1 was dysregulated in ESCC, and suppression of CRM1 expression which resulted in inhibiting of NF-κB signaling might be developed into a new strategy in ESCC therapy.
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Sigma-Aldrich
Fluorouracil, meets USP testing specifications
Sigma-Aldrich
Magnesium chloride solution, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, BioUltra, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, 2 M in H2O
Sigma-Aldrich
MISSION® esiRNA, targeting human XPO1
Supelco
5-Fluorouracil, analytical standard
Supelco
Fluorouracil, Pharmaceutical Secondary Standard; Certified Reference Material
Fluorouracil, European Pharmacopoeia (EP) Reference Standard
USP
Fluorouracil, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Xpo1
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Emb