Merck
  • Stabilization of chitosan/hyaluronan colloidal polyelectrolyte complexes in physiological conditions.
Stabilization of chitosan/hyaluronan colloidal polyelectrolyte complexes in physiological conditions.
Carbohydrate polymers (2015-01-08)
Danjun Wu, Thierry Delair
ABSTRACT
Polyelectrolyte complexes (PECs) between hyaluronan (HYA) and chitosan were obtained by the one-shot addition of default amounts of polyanion to an excess of polycation. The impact of intrinsic parameters (degree of polymerization and degree of acetylation) and extrinsic parameters (charge mixing ratio, the concentration and pH of polyelectrolyte solutions) on particle sizes and polydispersity were investigated. The PECs maintained their colloidal stability when stored in water. To preserve the colloidal stability at physiological salt concentration and pH, biological nontoxic metallic Zn(II) was added either post or during the formation of the particles. Dynamic light scattering results showed the PEC particle sizes in phosphate buffer saline remained constant and displayed a good stability at room temperature for at least 35 days, irrespective of the stabilization process by Zn(II). These results open promising prospects for the zinc cation stabilized chitosan-HYA PECs as efficient and safe tools for drug delivery.
Sigma-Aldrich
Sodium nitrite, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium nitrite, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥97%
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
Supelco
Sodium nitrite, analytical standard
Sigma-Aldrich
Sodium nitrite, ACS reagent, ≥97.0%
Millipore
Bifido Selective Supplement B, suitable for microbiology
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Supelco
Nitrite ion standard solution, 0.1 M NO2-, for ion-selective electrodes
Sigma-Aldrich
Sodium nitrite, 99.999% trace metals basis
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
Sodium nitrite, anhydrous, Redi-Dri, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC