Merck
  • Metabolites identification of glycyrin and glycyrol, bioactive coumarins from licorice.
Metabolites identification of glycyrin and glycyrol, bioactive coumarins from licorice.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-01-24)
Qi Wang, Xue Qiao, Yi Qian, Chun-fang Liu, Yan-fang Yang, Shuai Ji, Jun Li, De-an Guo, Min Ye
ABSTRACT
Coumarins are an important group of bioactive constituents in licorice (Glycyrrhiza uralensis), a worldwide popular herbal medicine. This study aims to elucidate the metabolism of two major licorice coumarins, glycyrin and glycyrol in rats. After oral administration of 40mg/kg glycyrin, neither the parent compound nor its metabolites could be detected in rats plasma or urine samples, indicating that glycyrin had poor oral bioavailability. Two hydroxylated metabolites, 4″-hydroxyl glycyrin and 5″-hydroxyl glycyrin, were detected in rat liver microsome incubation system. Among them, the major metabolite 4″-hydroxyl glycyrin, which is a new compound, was obtained by microbial transformation of Syncephalastrum racemosum AS 3.264. Its structure was fully identified by 1D and 2D NMR. Meanwhile, glycyrol, together with three metabolites, were detected in rats urine and fecal samples after oral administration (40mg/kg). Their structures were tentatively characterized by LC/MS. Glycyrol mainly undertakes hydroxylation metabolism, accompanied by hydration and dehydrogenation as minor reactions. This is the first systematic study on metabolism of glycyrin and glycyrol. The results could be valuable to evaluate druggability of these bioactive natural products.
Sigma-Aldrich
Dimethyl sulfoxide-d6, "Special HOH", ≥99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, anhydrous, 99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
1,3-Propylene sulfite, 99%
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.5 atom % D
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
D-Glucose 6-phosphate sodium salt, ≥98% (HPLC)
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%