Merck
In vitro release testing methods for vitamin E nanoemulsions.
International journal of pharmaceutics (2014-09-03)
Jacqueline M Morais, Diane J Burgess
ABSTRACT
This study reports the release properties of the poorly water-soluble active vitamin E acetate from oil/water nanoemulsions containing canola oil, CremophorRH40(®) and Span80(®) prepared using a low energy emulsification method (EPI process). Drug release was measured via dialysis sac and reverse dialysis sac methods as well as USP apparatus 4 fitted with dialysis sac adapters. Macro- and microscopic stability were evaluated and no instabilities were observed during the test period. In vitro release testing was adequately performed using the reverse dialysis sac and USP apparatus 4 methods. However, the dialysis method produced a slower release rate compared to the other methods and this was considered a result of violation of sink conditions within the dialysis sacs as well as inadequate mixing. Micellar solubilization was able to increase vitamin E transport from canola oil to buffer solution, but no model active concentration increase in the nanoemulsion external aqueous phase was observed despite the presence of micelles. Accordingly, it appeared that diffusion across the interfacial film was the rate-limiting step for in vitro release from these nanoemulsions. Sustained/prolonged release of vitamin E was observed and could be explained based on the high partition coefficient and on the nanoemulsion interfacial film proprieties.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Supelco
Tetrahydrofuran, analytical standard
Supelco
Tetrahydrofuran, Selectophore, ≥99.5%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Supelco
Tetrahydrofuran, HPLC grade, ≥99.9%, inhibitor-free
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Tetrahydrofuran, inhibitor-free, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Tetrahydrofuran, ReagentPlus®, ≥99.0%, contains 250 ppm BHT as inhibitor
Sigma-Aldrich
Tetrahydrofuran, contains 250 ppm BHT as inhibitor, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Supelco
Tetrahydrofuran, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Tetrahydrofuran, ACS reagent, ≥99.0%, contains 250 ppm BHT as inhibitor
Sigma-Aldrich
Tetrahydrofuran, contains 250 ppm BHT as inhibitor, ACS reagent, ≥99.0%
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium acetate trihydrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium acetate trihydrate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E262, 99.0-101.0% (calc. to the dried substance), ≤0.00002% Al
Sigma-Aldrich
Sodium acetate trihydrate, ACS reagent, ≥99%
Supelco
Span® 80, viscosity 1000-2000 mPa.s (20 °C)