Merck
Role of the focal adhesion protein TRIM15 in colon cancer development.
Biochimica et biophysica acta (2014-12-03)
Ok-Hee Lee, Jinkyoung Lee, Keun Ho Lee, Yun Mi Woo, Ju-Hee Kang, Ho-Geun Yoon, Soo-Kyung Bae, Zhou Songyang, Seung Hyun Oh, Youngsok Choi
ABSTRACT
The tripartite motif containing (TRIM) proteins are a large family of proteins that have been implicated in many biological processes including cell differentiation, apoptosis, transcriptional regulation, and signaling pathways. Here, we show that TRIM15 co-localized to focal adhesions through homo-dimerization and significantly suppressed cell migration. Domain mapping analysis indicated that B-box2 and PRY domains were essential for TRIM15 localization to focal adhesions and inhibition of cell migration. Our protein-protein interaction screen of TRIM15 with the integrin adhesome identified several TRIM15 interacting proteins including coronin 1B, cortactin, filamin binding LIM protein1, and vasodilator-stimulated phosphoprotein, which are involved in actin cytoskeleton dynamics. TRIM15 expression was tissue-restricted and downregulated in colon cancer. Level of TRIM15 expression was associated with colon cancer cell migration, as well as both in vitro and in vivo tumor growth. These data provide novel insights into the role of TRIM15 as an additional component of the integrin adhesome, regulating cell migration, and suggest that TRIM15 may function as a tumor suppressor of colon cancer.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
1-[2-(Trifluoromethyl)phenyl]imidazole
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride solution, 0.85%
SAFC
Sodium chloride solution, 5 M
Supelco
4-tert-Octylphenol monoethoxylate solution, 10 μg/mL in acetone, analytical standard
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Glutathione, European Pharmacopoeia (EP) Reference Standard
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
3,3′-Diaminobenzidine, 97%
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sucrose, Grade I, ≥99% (GC), suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture