Merck
  • Transient receptor potential channels and their role in modulating radial glial-neuronal interaction: a signaling pathway involving mGluR5.
Transient receptor potential channels and their role in modulating radial glial-neuronal interaction: a signaling pathway involving mGluR5.
Stem cells and development (2014-10-28)
Lauri M Louhivuori, Linda Jansson, Pauli M Turunen, Maria H Jäntti, Tommy Nordström, Verna Louhivuori, Karl E Åkerman
ABSTRACT
The guidance of developing neurons to the right position in the central nervous system is of central importance in brain development. Canonical transient receptor potential (TRPC) channels are thought to mediate turning responses of growth cones to guidance cues through fine control of calcium transients. Proliferating and 1- to 5-day-differentiated neural progenitor cells (NPCs) showed expression of Trpc1 and Trpc3 mRNA, while Trpc4-7 was not clearly detected. Time-lapse imaging showed that the motility pattern of neuronal cells was phasic with bursts of rapid movement (>60 μm/h), changes in direction, and intermittent slow phases or stallings (<40 μm/h), which frequently occurred in close contact with radial glial processes. Genetic interference with the TRPC3 and TRPC1 channel enhanced the motility of NPCs (burst frequency/stalling frequency). TRPC3-deficient cells or cells treated with the TRPC3 blocker pyr3 infrequently changed direction and seldom contacted radial glial processes. TRPC channels are also activated by group I metabotropic glutamate receptors (mGluR1 and mGluR5). As shown here, pyr3 blocked the calcium response mediated through mGluR5 in radial glial processes. Furthermore, 2-methyl-6-(phenylethynyl)pyridine, a blocker of mGluR5, affected the motility pattern in a similar way as TRPC3/6 double knockout or pyr3. The results suggest that radial glial cells exert attractant signals to migrating neuronal cells, which alter their motility pattern. Our results suggest that mGluR5 acting through TRPC3 is of central importance in radial glial-mediated neuronal guidance.
Sigma-Aldrich
Anti-MAP2A, 2B, 2C Antibody, clone HM-2, ascites fluid, clone HM-2, Chemicon®