Merck
  • The endogenous oxindole isatin induces apoptosis of MCF‑7 breast cancer cells through a mitochondrial pathway.
The endogenous oxindole isatin induces apoptosis of MCF‑7 breast cancer cells through a mitochondrial pathway.
Oncology reports (2014-09-02)
Zhongliang Ma, Lin Hou, Yuhong Jiang, Yanpin Chen, Jinlian Song
ABSTRACT
Isatin is an endogenous indole in mammalian tissues and fluids that is expected to have antitumor effects in human breast cancer cells. Human breast cancer cells (MCF-7) were exposed to isatin at various concentrations (0, 50, 100, 200 µmol/l) for 48 h. Apoptotic features were demonstrated by nuclei staining with Hoechst 33258 and flow cytometry. Bcl-2 and Bax mRNA were analyzed via reverse transcription-polymerase chain reaction. Bcl-2, Bax, the inhibitor of caspase-activated DNase (ICAD), and cytochrome c protein were analyzed by western blot analysis. Apoptosis, caspase-9 and -3 activation and mitochondrial depolarization were assayed by flow cytometry. The results showed that isatin induced apoptosis of MCF-7 cells. Furthermore, Bcl-2 expression was decreased and the ratio of Bcl-2 to Bax was significantly decreased by isatin. The mitochondrial transmembrane potential was markedly decreased and the release of cytochrome c into the cytosol was elevated following treatment with isatin. At the same time, caspase-9 and -3 were stimulated, followed by the degradation of ICAD, a caspase-3 substrate.
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Isatin, 97%
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Fluorescein, for fluorescence, free acid
Sigma-Aldrich
Propidium iodide solution
Fluorescein, European Pharmacopoeia (EP) Reference Standard