Merck
  • Determination of drugs of abuse in bovine dentin using liquid chromatography-electrospray ionization tandem mass spectrometry.
Determination of drugs of abuse in bovine dentin using liquid chromatography-electrospray ionization tandem mass spectrometry.
Journal of mass spectrometry : JMS (2014-12-06)
J Spinner, M Klima, J Kempf, L M Huppertz, V Auwärter, M J Altenburger, M A Neukamm
ABSTRACT
Drugs deposited in human teeth are well preserved; the spectrum of toxicological investigations may therefore be supplemented by an analysis method for drugs in teeth. A liquid chromatography-electrospray ionization tandem mass spectrometry assay for the detection and quantification of basic drugs of abuse in bovine dentin samples was developed and validated. The drugs and metabolites amphetamine, methamphetamine, methylenedioxymethylamphetamine, methylenedioxyethylamphetamine, codeine, morphine, cocaine and benzoylecgonine were extracted from 50 mg ground dentin powder by ultrasonication for 60 min in methanol 3 times. The extracts were analyzed on a triple-quadrupole mass-spectrometer in multiple reaction monitoring mode. The method was validated and proved to be accurate, precise, selective, specific and stable with good linearity within the calibration range and a lower limit of quantification of 10 to 20 pg/mg. To artificially load bovine dentin samples with drugs, the natural process of de- and remineralization in the oral cavity was mimicked by a pH-cycling experiment. The artificially drug-loaded dentin samples showed drug concentrations of 20 to 80 pg/mg. The method can be applied in further in vitro experiments as well as in post-mortem cases, especially where limited sample tissue is available.
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Potassium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Calcium chloride dihydrate, BioUltra, for molecular biology, ≥99.5% (KT)
Supelco
Calcium standard for AAS, analytical standard, 1.000 g/L Ca+2 in hydrochloric acid, traceable to BAM
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
Supelco
Potassium chloride solution, for Ag/AgCl electrodes, ~3 M KCl, saturated with silver chloride
Supelco
Potassium chloride solution, conductance standard B acc. to ISO 7888, 0.01 M KCl
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Calcium chloride dihydrate, tested according to Ph. Eur.
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Lactic acid, natural, ≥85%
Sigma-Aldrich
Lactic acid, 85%, FCC
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Potassium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Potassium chloride, tested according to Ph. Eur.
Supelco
Acetic acid, analytical standard
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Supelco
Potassium chloride solution, conductance standard C acc. to ISO 7888, 0.001 M KCl
Supelco
Potassium chloride solution, conductance standard A acc. to ISO 7888, 0.1 M KCl
Supelco
Potassium chloride solution, BioUltra, ~3 M in H2O
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
ISA (ionic strength adjustment solution: 1 M KCl), 1 M KCl
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Calcium chloride dihydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology