Merck
  • Tendon protein synthesis rate in classic Ehlers-Danlos patients can be stimulated with insulin-like growth factor-I.
Tendon protein synthesis rate in classic Ehlers-Danlos patients can be stimulated with insulin-like growth factor-I.
Journal of applied physiology (Bethesda, Md. : 1985) (2014-08-12)
Rie Harboe Nielsen, Lars Holm, Jacob Kildevang Jensen, Katja Maria Heinemeier, Lars Remvig, Michael Kjaer
ABSTRACT
The classic form of Ehlers-Danlos syndrome (cEDS) is an inherited connective tissue disorder, where mutations in type V collagen-encoding genes result in abnormal collagen fibrils. Thus the cEDS patients have pathological connective tissue morphology and low stiffness, but the rate of connective tissue protein turnover is unknown. We investigated whether cEDS affected the protein synthesis rate in skin and tendon, and whether this could be stimulated in tendon tissue with insulin-like growth factor-I (IGF-I). Five patients with cEDS and 10 healthy, matched controls (CTRL) were included. One patellar tendon of each participant was injected with 0.1 ml IGF-I (Increlex, Ipsen, 10 mg/ml) and the contralateral tendon with 0.1 ml isotonic saline as control. The injections were performed at both 24 and 6 h prior to tissue sampling. The fractional synthesis rate (FSR) of proteins in skin and tendon was measured with the stable isotope technique using a flood-primed continuous infusion over 6 h. After the infusion one skin biopsy and two tendon biopsies (one from each patellar tendon) were obtained. We found similar baseline FSR values in skin and tendon in the cEDS patients and controls [skin: 0.005 ± 0.002 (cEDS) and 0.007 ± 0.002 (CTRL); tendon: 0.008 ± 0.001 (cEDS) and 0.009 ± 0.002 (CTRL) %/h, mean ± SE]. IGF-I injections significantly increased FSR values in cEDS patients but not in controls (delta values: cEDS 0.007 ± 0.002, CTRL 0.001 ± 0.001%/h). In conclusion, baseline protein synthesis rates in connective tissue appeared normal in cEDS patients, and the patients responded with an increased tendon protein synthesis rate to IGF-I injections.
Sigma-Aldrich
tert-Butyldimethylsilyl chloride, reagent grade, 97%
Sigma-Aldrich
tert-Butyldimethylsilyl chloride solution, 1.0 M in THF
Sigma-Aldrich
IGF-I human, Animal-component free, recombinant, expressed in E. coli, suitable for cell culture
Sigma-Aldrich
tert-Butylchlorodimethylsilane solution, 50 wt. % in toluene