Merck
  • Peripheral biomarkers of oxidative stress and their limited potential in evaluation of clinical features of Huntington's patients.
Peripheral biomarkers of oxidative stress and their limited potential in evaluation of clinical features of Huntington's patients.
Biomarkers : biochemical indicators of exposure, response, and susceptibility to chemicals (2014-07-02)
Irene Ciancarelli, Daniela De Amicis, Caterina Di Massimo, Carla Di Scanno, Caterina Pistarini, Nicolantonio D'Orazio, Maria Giuliana Tozzi Ciancarelli
ABSTRACT
Peripheral oxidative biomarkers could be useful for monitoring clinical features of Huntington's disease (HD). Cu/Zn-superoxide dismutase (Cu/Zn-SOD), neuron-specific enolase (NSE) and 8-hydroxy-2'-deoxyguanosine (8-oxoGua) serum levels were analysed in 18 HD patients and 10 controls. Clinical measures were recorded from each HD patients. Cu/Zn-SOD, NSE and 8-oxoGua values were higher in HD patients than in controls. Cu/Zn-SOD and NSE correlated positively. No correlation was observed between the biomarkers analysed and the clinical measures assessed. Serum oxidative biomarkers could express the neuronal oxidative processes going on in HD patients but are inadequate to evaluate clinical features of the disease.
Sigma-Aldrich
Human Thrombopoietin ELISA Kit, for serum, plasma, cell culture supernatants and urine
Sigma-Aldrich
Mouse Thrombopoietin ELISA Kit, for serum, plasma and cell culture supernatant