Merck
  • Regulation of cell survival by HUNK mediates breast cancer resistance to HER2 inhibitors.
Regulation of cell survival by HUNK mediates breast cancer resistance to HER2 inhibitors.
Breast cancer research and treatment (2014-12-18)
Elizabeth S Yeh, Melissa A Abt, Elizabeth G Hill
ABSTRACT
Breast cancer patients who are HER2-positive receive targeted inhibitors to HER2, including trastuzumab and lapatinib. While patients benefit from the use of HER2 inhibitors, many fail therapy and almost all patients become resistant to treatment, indicating a critical need to prevent treatment failure. Several recent studies indicate that activation of autophagy contributes to trastuzumab and lapatinib resistance and demonstrate that impairing autophagy in breast cancer cells is therapeutically beneficial. Moreover, autophagy is mechanistically linked through signaling crosstalk to apoptotic pathways, where activation of one process impacts the other. Therefore, understanding the molecular mechanisms that control these processes may uncover novel areas of therapeutic intervention to combat or prevent resistance in breast cancer. We previously characterized the protein kinase HUNK as a breast cancer-promoting factor in HER2/neu-induced mammary tumor models, in which HUNK supported the survival of HER2/neu-positive tumor cells, likely through the regulation of apoptosis. Because significant crosstalk exists between apoptotic and autophagy proteins, we now examine if HUNK is also able to regulate cell survival through modulation of autophagy using HER2 inhibitor sensitive and resistant breast cancer models. Furthermore, we investigate whether inhibiting HUNK impairs in vivo tumor growth that is initiated by HER2 inhibitor-resistant breast cancer cells. Our findings indicate that therapeutically targeting HUNK is a potential strategy for overcoming resistance and that resistant breast cancer cells maintain HUNK expression to drive tumorigenesis, an observation that is consistent with a pro-survival role for this kinase.
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
MISSION® esiRNA, targeting human HUNK
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Trypan Blue, ≥80% (HPLC), Dye content 60 %
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O