Merck
  • Pharmacokinetics and tissue distribution of ginsenoside Rh2 and Rg3 epimers after oral administration of BST204, a purified ginseng dry extract, in rats.
Pharmacokinetics and tissue distribution of ginsenoside Rh2 and Rg3 epimers after oral administration of BST204, a purified ginseng dry extract, in rats.
Xenobiotica; the fate of foreign compounds in biological systems (2014-06-17)
Soo Hyeon Bae, Jung Bae Park, Yu Fen Zheng, Min Jung Jang, Sun Ok Kim, Jeom Yong Kim, Young Hyo Yoo, Kee Dong Yoon, Euichaul Oh, Soo Kyung Bae
ABSTRACT
1. BST204, a purified ginseng dry extract containing a high concentration of racemic Rh2 and Rg3 mixtures, is being developed for supportive care use in cancer patients in Korea. This study investigates the pharmacokinetics and tissue distribution of BST204 in rats. 2. After oral administration of BST204, only the S epimers, S-Rh2 and S-Rg3, could be determined in rat plasma. The poor absorption of the R-epimers, R-Rh2 and R-Rg3, may be attributed to lower membrane permeability and extensive intestinal oxygenation and/or deglycosylation into metabolites. The AUC and Cmax values of both S-Rh2 and S-Rg3 after BST204 oral administration were proportional to the administered BST204 doses ranged from 400 mg/kg to 2000 mg/kg, which suggested linear pharmacokinetic properties. 3. There were no statistically significant differences in the pharmacokinetics of S-Rh2 and S-Rg3 after oral administration of pure S-Rh2 (31.5 mg/kg) and S-Rg3 (68 mg/kg) compared with oral administration of BST204, 1000 mg/kg. These indicated that the presence of other components of BST204 extract did not influence the pharmacokinetic behavior of S-Rh2 and S-Rg3. 4. After oral dosing of BST204, S-Rh2 and S-Rg3 were distributed mainly to the liver and gastrointestinal tract in rats. 5. Our finding may help to understand pharmacokinetic characteristics of S-Rh2, R-Rh2, S-Rg3, and R-Rg3, comprehensively, and provide useful information in clinical application of BST204.
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%