Merck
Fluorine in drug design: a case study with fluoroanisoles.
ChemMedChem (2015-03-11)
Li Xing, David C Blakemore, Arjun Narayanan, Ray Unwalla, Frank Lovering, R Aldrin Denny, Huanyu Zhou, Mark E Bunnage
ABSTRACT
Anisole and fluoroanisoles display distinct conformational preferences, as evident from a survey of their crystal structures. In addition to altering the free ligand conformation, various degrees of fluorination have a strong impact on physicochemical and pharmacokinetic properties. Analysis of anisole and fluoroanisole matched molecular pairs in the Pfizer corporate database reveals interesting trends: 1) PhOCF3 increases log D by ~1 log unit over PhOCH3 compounds; 2) PhOCF3 shows lower passive permeability despite its higher lipophilicity; and 3) PhOCF3 does not appreciably improve metabolic stability over PhOCH3 . Emerging from the investigation, difluoroanisole (PhOCF2 H) strikes a better balance of properties with noticeable advantages of log D and transcellular permeability over PhOCF3 . Synthetic assessment illustrates that the routes to access difluoroanisoles are often more straightforward than those for trifluoroanisoles. Whereas replacing PhOCH3 with PhOCF3 is a common tactic to optimize ADME properties, our analysis suggests PhOCF2 H may be a more attractive alternative, and greater exploitation of this motif is recommended.
Product Description
Supelco
1-Octanol, analytical standard
Sigma-Aldrich
1-Octanol, natural, ≥98%, FCC
Sigma-Aldrich
1-Octanol, anhydrous, ≥99%
Sigma-Aldrich
1-Octanol, ≥98%, FCC, FG
Sigma-Aldrich
1-Octanol, suitable for HPLC, ≥99%
Sigma-Aldrich
1-Octanol, ReagentPlus®, 99%
Sigma-Aldrich
1-Octanol, ACS reagent, ≥99%
Sigma-Aldrich
Anisole, ≥99%, FCC, FG
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Supelco
Dimethyl sulfoxide, analytical standard
Supelco
Anisole, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Anisole, anhydrous, 99.7%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Anisole, ReagentPlus®, 99%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Supelco
Anisole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)