Merck
  • Heme oxygenase-1 (HO-1) mediated respiratory responses to hypoxia in the goldfish, Carassius auratus.
Heme oxygenase-1 (HO-1) mediated respiratory responses to hypoxia in the goldfish, Carassius auratus.
Respiratory physiology & neurobiology (2014-05-02)
Velislava Tzaneva, Steve F Perry
ABSTRACT
In this study we investigated the role of heme oxygenase-1 (HO-1) in modulating the hypoxic and hyperoxic ventilatory responses of goldfish (Carassius auratus) acclimated to 7 and 25°C. HO-1 was present in the neuroepithelial cells (NECs; putative branchial O2 chemoreceptors) of fish acclimated to 7°C only. Hypoxia exposure increased gill HO-1 activity in 7°C fish (14.0±1.4 to 42.5±3.2pmolbilirubinmin(-1)mgprotein(-1)). Inhibition of HO-1 activity with zinc protophorphyrin IX (ZnPPIX) increased the ventilation frequency response to acute hypoxia (30mmHg); frequency increased from 48.3±5.1 to 137.4±16.0 breaths per min (BPM) in hypoxic 7°C fish treated with ZnPPIX compared to 46.2±4.2 to 77.9±5.3 BPM in control fish. Unlike in the control (untreated) 7°C fish exposed to hyperoxia, fish injected with ZnPPIX did not significantly decrease breathing frequency. Inhibiting HO-1 activity was without effect on the hypoxic or hyperoxic ventilatory responses of fish acclimated to 25°C. Based on these observations, we suggest that HO-1 plays an inhibitory role in regulating breathing frequency but only in goldfish acclimated to 7°C.
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Benzocaine, ≥99% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Supelco
Benzocaine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Benzocaine, European Pharmacopoeia (EP) Reference Standard
USP
Benzocaine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Bilirubin, purum, ≥95.0% (UV)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
Ethyl 4-aminobenzoate, 98%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture