Merck
  • Hydrogen from formic acid through its selective disproportionation over sodium germanate--a non-transition-metal catalysis system.
Hydrogen from formic acid through its selective disproportionation over sodium germanate--a non-transition-metal catalysis system.
Angewandte Chemie (International ed. in English) (2014-08-30)
Ruth I J Amos, Falk Heinroth, Bun Chan, Sisi Zheng, Brian S Haynes, Christopher J Easton, Anthony F Masters, Leo Radom, Thomas Maschmeyer
ABSTRACT
A robust catalyst for the selective dehydrogenation of formic acid to liberate hydrogen gas has been designed computationally, and also successfully demonstrated experimentally. This is the first such catalyst not based on transition metals, and it exhibits very encouraging performance. It represents an important step towards the use of renewable formic acid as a hydrogen-storage and transport vector in fuel and energy applications.
Sigma-Aldrich
Germanium(IV) oxide, (crystalline powder), 99.998% trace metals basis
Sigma-Aldrich
Germanium(IV) oxide, ≥99.99% trace metals basis
Sigma-Aldrich
Tetrapropylammonium hydroxide solution, 1.0 M in H2O