Merck
  • Transcriptomic profiling explains racial disparities in pterygium patients treated with doxycycline.
Transcriptomic profiling explains racial disparities in pterygium patients treated with doxycycline.
Investigative ophthalmology & visual science (2014-11-02)
Ignacio M Larrayoz, Óscar Rúa, Sara Velilla, Alfredo Martínez
ABSTRACT
To understand the differential responses to doxycycline between Caucasian and Hispanic patients observed in a previous clinical trial. Primary cultures were established using pterygia excised from male Caucasian (n = 3) and Hispanic (n = 6) patients. The response of these cells to doxycycline was tested in a toxicity assay. In addition, a complete transcriptome was obtained from the nine samples, and the results were analyzed using false discovery rate statistics. Results were confirmed by quantitative RT (qRT)-PCR and Western blotting for a limited set of genes. Caucasian pterygium cells underwent apoptosis upon exposure to doxycycline, whereas Hispanic cells survived the treatment. Transcriptomic analysis showed profound differences between cells of both ethnicities, even before treatment, implicating important cellular pathways such as the mitochondrial oxidative phosphorylation chain, the proteasome, and the components of the extracellular matrix. Following exposure to doxycycline, there was a significant increase in proapoptotic proteins, regulators of the cell cycle, and components of the mitochondrial membrane in Caucasian cells but not in their Hispanic counterparts. There was a good correlation between data obtained by ultrasequencing and those generated by qRT-PCR or Western blotting. The lack of response to doxycycline observed in Hispanic pterygium patients in a previous clinical trial can be explained by the genetic protection afforded to the cells in this ethnic background against apoptosis and cell death. New therapeutic options must be devised for these patients.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent