Merck
  • Fast determination of catechins and xanthines in tea beverages by micellar electrokinetic chromatography.
Fast determination of catechins and xanthines in tea beverages by micellar electrokinetic chromatography.
Journal of agricultural and food chemistry (2003-02-20)
Matteo Bonoli, Piero Colabufalo, Marco Pelillo, Tullia Gallina Toschi, Giovanni Lercker
ABSTRACT
Antioxidant properties and stimulating effects of green tea are related to its content of cathechins and xanthines; tea quality evaluation is based on organoleptic tests and on the presence of those components. In this work, by a MEKC method, eight cathechins and three xanthines were quantified in some tea-based beverages. The best separation was realized using a phosphate-borate running buffer, with sodium dodecyl sulfate as micellar agent. A 40 cm capillary, a temperature of 29 degrees C, a voltage of 30 kV, and UV detection at 200 nm were used. The method showed a very good sensitivity (limit of detection ranging from 0.0011 to 0.0051 microg/mL) and was applied to real tea samples to characterize their antioxidant content. Statistical studies were performed and showed a satisfactory reliability of the data.
Product Description
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Pricing and availability is not currently available.