Merck
  • The synthesis of internal standards for the quantitative determination of sphingolipids by tandem mass spectrometry.
The synthesis of internal standards for the quantitative determination of sphingolipids by tandem mass spectrometry.
Rapid communications in mass spectrometry : RCM (2005-05-24)
Kevin Mills, Simon Eaton, Victoria Ledger, Elisabeth Young, Bryan Winchester
ABSTRACT
Novel internal standards have been synthesised for the quantitative determination by tandem mass spectrometry (MS/MS) of the sphingolipids that accumulate in lysosomal storage diseases. The [d4]C16- and [d47]C24-isoforms of galactosylceramide (CMH), lactosylceramide (CDH), globotriaosylceramide (CTH), cerebroside sulphate, sphingomyelin and G(M1)-, G(M2)- and G(M3)-gangliosides were synthesised by the reaction of their lyso-forms with the acid chlorides of hexadecanoic 5,5,6,6-d4 acid ([d4]-palmitic acid) and tetracosanoic-d47 acid ([d47]-lignoceric acid), respectively. The acid chlorides were formed using oxalyl chloride. The structures of the internal standards were confirmed by MS/MS. The fragmentation pattern of each novel compound was similar to that of the corresponding natural form of the sphingolipid, making it a good internal standard for the quantitative determination of the natural sphingolipid by ESI-MS/MS. Characteristic product ions were identified for each compound.