Merck
  • A checkpoint protein that scans the chromosome for damage at the start of sporulation in Bacillus subtilis.
A checkpoint protein that scans the chromosome for damage at the start of sporulation in Bacillus subtilis.
Cell (2006-05-23)
Michal Bejerano-Sagie, Yaara Oppenheimer-Shaanan, Idit Berlatzky, Alex Rouvinski, Mor Meyerovich, Sigal Ben-Yehuda
ABSTRACT
In response to DNA damage, cells activate checkpoint signaling cascades to control cell-cycle progression and elicit DNA repair in order to maintain genomic integrity. The sensing and repair of lesions is critical for Bacillus subtilis cells entering the developmental process of sporulation as damaged DNA may prevent the cells from completing spore morphogenesis. We report the identification of the protein DisA (DNA integrity scanning protein, annotated YacK), which is required to delay the initiation of sporulation in response to chromosomal damage. DisA is a nonspecific DNA binding protein that forms a single focus, which moves rapidly within the bacterial cell, pausing at sites of DNA damage. We propose that the DisA focus scans along the chromosomes searching for lesions. Upon encountering a lesion, DisA delays entry into sporulation until the damage is repaired.
Sigma-Aldrich
DAPI, for nucleic acid staining
Pricing and availability is not currently available.