Merck
  • Forward mutations and DNA-protein crosslinks induced by ammonium metavanadate in cultured mammalian cells.
Forward mutations and DNA-protein crosslinks induced by ammonium metavanadate in cultured mammalian cells.
Mutation research (1992-09-01)
M D Cohen, C B Klein, M Costa
ABSTRACT
Ammonium metavanadate yielded a dose-dependent increase in mutation frequency at the V79 hprt locus following a 24-h exposure period in serum-free F12 medium. Vanadate also increased the mutation frequency of V79 cells by exposure of cells in salts-glucose medium, but these effects were not as striking, or as dose-dependent as they were in serum-free F12 medium. Ammonium metavanadate enhanced the mutation frequency in a V79 variant containing a transfected bacterial gpt gene. These cells are known to be more responsive to oxidative type mutations, and to mutations involving deletions. Although the absolute level of mutations was greater in these cells with ammonium metavanadate, so was the background, and these cells did not exhibit an enhanced mutagenic response to vanadate when compared to the wild-type V79 cells. The vanadate results were compared to a positive control potassium chromate, which exhibited a dose-dependent increase in mutation frequency. Ammonium metavanadate induced DNA-protein crosslinks formation in both Chinese hamster ovary and human MOLT4 cells, and the role of these relatively unrepaired genetic lesions in the mutations produced by vanadate and chromate are discussed.
Sigma-Aldrich
Ammonium metavanadate, puriss. p.a., ACS reagent, ≥99.0% (RT)