Merck
  • Identification of the human ubiquitin specific protease 31 (USP31) gene: structure, sequence and expression analysis.
Identification of the human ubiquitin specific protease 31 (USP31) gene: structure, sequence and expression analysis.
DNA sequence : the journal of DNA sequencing and mapping (2004-09-10)
Paul J Lockhart, Mary Hulihan, Sarah Lincoln, Jennifer Hussey, Lisa Skipper, Gina Bisceglio, Kristen Wilkes, Matthew J Farrer
ABSTRACT
Parkinson's disease (PD) is a common neurodegenerative disorder with clinical features of bradykinesia, rigidity and resting tremor resulting from the deficiency of dopamine in the nigrostriatal system. Recently, PARK6 was identified as a novel locus associated with autosomal recessive PD. Here we report the identification and characterization of a novel human deubiquitylating gene (USP31), which maps to the critical PARK6 region. Database analysis and 5' RACE identified a 4070bp cDNA, encoded by 27 exons spanning approximately 105kbp of genomic sequence. The predicted protein of 1035 amino acids included a conserved ubiquitin hydrolase region (Prosite profile PS50235), a DUSP (domain in ubiquitin specific proteases-Smart00695) and a ubiquitin-like domain (Prosite pattern PS00299). Northern blot analysis revealed a single USP31 transcript of approximately 4 kb, which was primarily expressed in the testis and lung.