- Efficient palladium-catalyzed N-arylation of indoles.
Efficient palladium-catalyzed N-arylation of indoles.
Organic letters (2000-05-18)
D W Old, M C Harris, S L Buchwald
PMID10814458
ABSTRACT
[equation--see text] The N-arylation of indoles, including a variety of substituted ones, has been carried out using bulky, electron-rich phosphines as the supporting ligand in combination with Pd(2)(dba)(3). Using this catalyst system, the efficient coupling of indole and a variety of substituted indoles with aryl iodides, bromides, chlorides, and triflates can be achieved.