Merck
  • Multi-target-directed coumarin derivatives: hAChE and BACE1 inhibitors as potential anti-Alzheimer compounds.
Multi-target-directed coumarin derivatives: hAChE and BACE1 inhibitors as potential anti-Alzheimer compounds.
Bioorganic & medicinal chemistry letters (2007-11-14)
Lorna Piazzi, Andrea Cavalli, Francesco Colizzi, Federica Belluti, Manuela Bartolini, Francesca Mancini, Maurizio Recanatini, Vincenza Andrisano, Angela Rampa
ABSTRACT
The complex etiology of Alzheimer's disease (AD) prompts scientists to develop multifunctional compounds to combat causes and symptoms of such neurodegeneration. To this aim we designed, synthesized, and tested a series of compounds by introducing halophenylalkylamidic functions on the scaffold of AP2238, which is a dual binding site acetylcholinesterase inhibitor. The inhibitory activity was successfully extended to the beta-site amyloid precursor protein cleavage enzyme, leading to the discovery of a potent inhibitor of this enzyme (3) and affording multifunctional compounds (2, 6, 8) for the treatment of AD.