Merck
  • Hepatitis B virus large envelope protein interacts with gamma2-adaptin, a clathrin adaptor-related protein.
Hepatitis B virus large envelope protein interacts with gamma2-adaptin, a clathrin adaptor-related protein.
Journal of virology (2001-05-03)
C Hartmann-Stühler, R Prange
ABSTRACT
For the outcome of a hepatitis B virus (HBV) infection, the viral L envelope protein with its pre-S domain performs pivotal functions by mediating attachment of HBV to liver cells, envelopment of viral capsids, release of (sub)viral particles, regulation of supercoiled DNA amplification, and transcriptional transactivation. To assess its multiple functions and host-protein assistance involved, we initiated a two-hybrid screen using the L-specific pre-S1 domain as bait. With this approach, we have identified gamma2-adaptin, a putative member of the clathrin adaptor proteins responsible for protein sorting and trafficking, as a specific binding partner of L protein. Evidence for a physical interaction between L protein and gamma2-adaptin was also demonstrated by affinity chromatography and coimmunoprecipitation, and the binding sites were mapped to the L-specific pre-S1 domain and the gamma2-adaptin-specific ear domain. The specificity of the interaction was further sustained by the failure of gamma1-adaptin, a closely related gamma2-adaptin homologue, to associate with L protein. Analysis of an L mutant protein indicates that the L-gamma2-adaptin interaction strictly depends on the pre-S1 domain of transmembrane L protein oriented to the cytosol and thus appears to occur in the cytosolic environment. Interestingly, coexpression of the two interacting partners in transfected cells resulted in recruitment of gamma2-adaptin by L protein onto cis-Golgi-like structures, strongly indicating that the association is physiologically relevant. Together, the results suggest a role for gamma2-adaptin in L-mediated processes of viral biogenesis and/or pathogenesis, such as facilitating and guiding HBV assembly.