- Ugi/Robinson-Gabriel reactions directed toward the synthesis of 2,4,5-trisubstituted oxazoles.
Ugi/Robinson-Gabriel reactions directed toward the synthesis of 2,4,5-trisubstituted oxazoles.
Tetrahedron letters (2013-04-06)
Arthur Y Shaw, Zhigang Xu, Christopher Hulme
PMID23559684
ABSTRACT
This Letter discloses a novel concise synthesis of a series of 2,4,5-trisubstituted oxazoles via a tandem Ugi/Robinson-Gabriel sequence. Herein, 2,4-dimethoxybenzylamine