Merck
  • Evaluation of thyroid hormone levels in chronic kidney disease patients.
Evaluation of thyroid hormone levels in chronic kidney disease patients.
Saudi journal of kidney diseases and transplantation : an official publication of the Saudi Center for Organ Transplantation, Saudi Arabia (2015-01-13)
Gandham Rajeev, Wilma Delphine Silvia Chickballapur Rayappa, Ravella Vijayalakshmi, Manchala Swathi, Sunil Kumar
ABSTRACT
We attempted in this study to determine the thyroid hormone levels in 45 adult chronic kidney disease (CKD) patients and 45 ageand sex-matched healthy subjects as controls. The serum thyroid hormone levels were measured by a radioimmunoassay. Serum concentrations of creatinine, urea, electrolytes and total proteins and albumin were measured as well. There was a significant decrease in the levels of serum total T3, total T4 and total protein and albumin levels in CKD patients when compared with the controls. There was a significant increase in the level of thyroid stimulating hormone in the CKD patients compared with the controls. Our study suggests that CKD leads to significant changes in the thyroid hormone levels, which need to be interpreted carefully in these patients.
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
L-Thyroxine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Thyrotropic hormone from bovine pituitary
Sigma-Aldrich
L-Thyroxine, ≥98% (HPLC)
Sigma-Aldrich
Thyrotropic hormone from human pituitary, lyophilized powder, ≥6.2 IU/mg (immunoassay, non-sterile)
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, BioUltra, for molecular biology, 99% (T)
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Supelco
L-Thyroxine (T4) solution, 100 μg/mL in methanol with 0.1N NH3, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Urea, European Pharmacopoeia (EP) Reference Standard
Supelco
Urea, analytical standard
USP
Levothyroxine, United States Pharmacopeia (USP) Reference Standard
USP
Urea, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Urea, suitable for electrophoresis