Merck
  • Response of aerobic granular sludge to the long-term presence to nanosilver in sequencing batch reactors: reactor performance, sludge property, microbial activity and community.
Response of aerobic granular sludge to the long-term presence to nanosilver in sequencing batch reactors: reactor performance, sludge property, microbial activity and community.
The Science of the total environment (2014-12-03)
Xiangchun Quan, Yan Cen, Fang Lu, Lingyun Gu, Jingyun Ma
ABSTRACT
The increasing use of silver nanoparticles (Ag NPs) raises concerns about their potential toxic effects on the environment. Granular shape sludge is a special type of microbial aggregate. The response of aerobic granular sludge (AGS) to the long-term presence of Ag NPs has not been well studied. In this study, AGS was exposed to 5 and 50mg/L Ag NPs in sequence batch reactors (SBRs) for 69 days, and its response was evaluated based on the sludge properties, microbial activity and community, and reactor performance. The results showed that Ag NPs caused inhibition to microbial activities of AGS from Day 35. At the end of 69 days of Ag NPs exposure, the microbial activity of AGS was significantly inhibited in terms of inhibitions of the ammonia oxidizing rate (33.0%), respiration rate (17.7% and 45.6%) and denitrification rate (6.8%), as well as decreases in the ammonia mono-oxygenase and nitrate reductase activities. During the long-term exposure, the AGS maintained its granular shape and large granule size (approximately 900 μm); the microbial community of AGS slightly changed, but the dominant microbial population remained. Overall, the AGS tolerated the toxicity of Ag NPs well, but a long-term exposure may produce chronic toxicity to the AGS, which is concerning.
Sigma-Aldrich
Silver nanowires, diam. × L 100 nm × 6 μm, 0.5% (isopropyl alcohol suspension)
Sigma-Aldrich
Silver, conductive paste
Sigma-Aldrich
Silver, wire, diam. 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Silver, powder, <250 μm, 99.99% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 0.1 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, colloidal, 65-75% Ag basis
Sigma-Aldrich
Silver, wool, diam. 0.05 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 1.0 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 2.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver nanowires, diam. × L 200  nm × 10 μm, 0.5% (isopropyl alcohol suspension)
Silver, insulated wire, 2m, conductor diameter 0.025mm, insulation thickness 0.005mm, polyester insulation, 99.99%
Silver, sphere, 100pcs, diameter 0.5mm, precision sphere grade
Silver, tube, 200mm, outside diameter 3.5mm, inside diameter 3.04mm, wall thickness 0.23mm, as drawn, 99.95+%
Silver, tube, 100mm, outside diameter 0.7mm, inside diameter 0.3mm, wall thickness 0.2mm, as drawn, 99.95+%
Silver, microfoil, disks, 25mm, thinness 0.5μm, specific density 522.6μg/cm2, permanent mylar 3.5μm support, 99.95+%
Silver, insulated wire, 10m, conductor diameter 0.025mm, insulation thickness 0.005mm, polyurethane insulation, 99.99%
Silver, insulated wire, 50m, conductor diameter 0.25mm, insulation thickness 0.024mm, PTFE (polytetrafluoroethylene) insulation, 99.99%
Silver, insulated wire, 2m, conductor diameter 0.25mm, insulation thickness 0.024mm, PTFE (polytetrafluoroethylene) insulation, 99.99%
Silver, insulated wire, 5m, conductor diameter 0.25mm, insulation thickness 0.015mm, epoxy insulation, 99.99%
Silver, rod, 500mm, diameter 3.0mm, as drawn, 99.997%
Silver, insulated wire, 5m, conductor diameter 0.025mm, insulation thickness 0.005mm, polyester insulation, 99.99%
Silver, rod, 500mm, diameter 3.5mm, as drawn, 99.95+%
Silver, insulated wire, 2m, conductor diameter 0.025mm, insulation thickness 0.005mm, polyurethane insulation, 99.99%
Silver, sphere, 200pcs, diameter 0.5mm, precision sphere grade
Silver, rod, 100mm, diameter 8.0mm, as drawn, 99.95+%
Silver, tube, 200mm, outside diameter 10mm, inside diameter 8mm, wall thickness 1.0mm, as drawn, 99.95+%
Silver, insulated wire, 50m, conductor diameter 0.025mm, insulation thickness 0.005mm, polyester insulation, 99.99%