Merck
  • Enrichment of reticulocytes from whole blood using aqueous multiphase systems of polymers.
Enrichment of reticulocytes from whole blood using aqueous multiphase systems of polymers.
American journal of hematology (2014-09-30)
Ashok A Kumar, Caeul Lim, Yovany Moreno, Charles R Mace, Abeer Syed, Daria Van Tyne, Dyann F Wirth, Manoj T Duraisingh, George M Whitesides
ABSTRACT
This paper demonstrates the enrichment of reticulocytes by centrifuging whole blood through aqueous multiphase systems (AMPSs)-immiscible phases of solutions of polymers that form step-gradients in density. The interfaces of an AMPS concentrate cells; this concentration facilitates the extraction of blood enriched for reticulocytes. AMPS enrich reticulocytes from blood from both healthy and hemochromatosis donors. Varying the osmolality and density of the phases of AMPS provides different levels of enrichment and yield of reticulocytes. A maximum enrichment of reticulocytemia of 64 ± 3% was obtained from donors with hemochromatosis. When used on peripheral blood from normal donors, AMPS can provide a higher yield of enriched reticulocytes and a higher proportion of reticulocytes expressing CD71 than differential centrifugation followed by centrifugation over Percoll. Blood enriched for reticulocytes by AMPS could be useful for research on malaria. Several species of malaria parasites show a preference to invade young erythrocytes and reticulocytes; this preference complicates in vitro cultivation of these species in human blood. Plasmodium knowlesi malaria parasites invade normal human blood enriched for reticulocytes by AMPSs at a rate 2.2 times greater (P < 0.01) than they invade unenriched blood. Parasite invasion in normal blood enriched by AMPS was 1.8 times greater (P < 0.05) than in blood enriched to a similar reticulocytemia by differential centrifugation followed by centrifugation over Percoll. The enrichment of reticulocytes that are invaded by malaria parasites demonstrates that AMPSs can provide a label-free method to enrich cells for biological research.
Sigma-Aldrich
Mowiol® 6-98, Mw ~47,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 410,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 670,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 25,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, Mr ~200,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 12,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~70,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr 450,000-650,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, Mr ~60,000
Supelco
Dextran, analytical standard, for GPC, Set Mp 1,000-400,000
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Poly(vinyl alcohol), Mw 146,000-186,000, 99+% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 31,000-50,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 13,000-23,000, 98% hydrolyzed
Supelco
Dextran, analytical standard, for GPC, 12,000
Supelco
Dextran, analytical standard, for GPC, 25,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr 15,000-25,000
Supelco
Dextran, analytical standard, for GPC, 410,000
Supelco
Dextran, analytical standard, for GPC, 270,000
Sigma-Aldrich
Mowiol® 4-88, Mw ~31,000
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 130,000, 99+% hydrolyzed
Sigma-Aldrich
Mowiol® 8-88, Mw ~67,000
Sigma-Aldrich
Mowiol® 10-98, Mw ~61,000
Sigma-Aldrich
Poly(vinyl alcohol), Mw 13,000-23,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 85,000-124,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 146,000-186,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 31,000-50,000, 98-99% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 89,000-98,000, 99+% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 85,000-124,000, 99+% hydrolyzed