Merck
  • Identification and characterization of the aroma-impact components of Thai fish sauce.
Identification and characterization of the aroma-impact components of Thai fish sauce.
Journal of agricultural and food chemistry (2015-03-03)
Nawaporn Lapsongphon, Jirawat Yongsawatdigul, Keith R Cadwallader
ABSTRACT
Comprehensive analysis of the potent odorants in Thai premium fish sauce samples was accomplished by use of complementary volatile isolation methods combined with gas chromatography-olfactometry (GC-O) and GC-mass spectrometry. Odorants of intermediate and low volatility were determined by direct solvent extraction/solvent-assisted flavor evaporation (DSE-SAFE) and aroma extract dilution analysis (AEDA). Meanwhile, static headspace dilution analysis (SHDA) and headspace solid-phase microextraction (H-SPME) were used to determine the highly volatile odorants. Results of AEDA indicated the importance (log3FD factor≥6) of five acidic odorants (butanoic acid, 3-methylbutanoic acid, 4-hydroxy-2,5-dimethyl-3(2H)-furanone, 4-hydroxy-2-ethyl-5-methyl-3(2H)-furanone, and 2-phenylacetic acid) and four neutral/basic odorants (3-methylbutanal, (Z)-1,5-octadien-3-one, phenylacetaldehyde, and o-aminoacetophone). Results of SHDA indicated the predominant (log3FD factors≥5) headspace odorants were methanethiol, 2-methylpropanal, 2-methylbutanal, 3-methylbutanal, dimethyl trisulfide, 3-(methylthio)propanal, and butanoic acid. Concentrations for 21 odorants were determined by stable isotope dilution analysis (SIDA), and their odor-activity values (OAVs) were calculated. Among these, methanethiol, 2-methylpropanal, 3-methylbutanal, dimethyl trisulfide, 3-(methylthio)propanal, and butanoic acid had the highest OAVs (>500). Results of aroma recombination and omission studies revealed the importance of acids, aldehydes, and sulfur-containing compounds to the overall aroma of the Thai fish sauce.
Product Description
Supelco
SPME Sampling Stand, for use with 4 mL vials
Supelco
SPME Sampling Stand, for use with 15 mL vials
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 30 μm(PDMS, needle size 24 ga, for use with autosampler
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 30 μm(PDMS, needle size 24 ga, for use with manual holder
Supelco
SPME fiber assembly, Carbowax-Polyethylene Glycol (PEG) Coating, needle size 23 ga, df 60 μm(PEG, for use with autosampler
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 23 ga, PDMS/DVB StableFlex, for use with autosampler
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm(PDMS, needle size 23 ga, for use with autosampler
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 24 ga, StableFlex, for use with manual holder
Supelco
SPME Portable Field Sampler, coating PDMS/DVB
Supelco
SPME fiber assembly polyacrylate (PA), df 85 μm(PA, for use with autosampler, needle size 24 ga
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, for use with manual holder, needle size 23 ga
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 24 ga, StableFlex, for use with autosampler
Supelco
SPME fiber assembly, Carbowax-Polyethylene Glycol (PEG) Coating, needle size 23 ga, df 60 μm(PEG, metal alloy fiber, for use with manual holder
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, needle size 24 ga, for use with manual holder
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm(PDMS, needle size 24 ga, for use with autosampler
Supelco
SPME Portable Field Sampler, coating CAR/PDMS
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, for use with autosampler, needle size 23 ga
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, for use with autosampler, needle size 24 ga
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, for use with autosampler, needle size 23 ga
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), fused silica fiber, df 65 μm(PDMS/DVB, for use with manual holder, needle size 23 ga
Supelco
SPME fiber assembly polyacrylate (PA), df 85 μm(PA, for use with autosampler, needle size 23 ga
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 24 ga, for use with manual holder
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), metal alloy fiber, df 100 μm(PDMS, needle size 23 ga, for use with autosampler
Supelco
SPME fiber assembly polyacrylate (PA), df 85 μm(PA, for use with manual holder, needle size 24 ga
Supelco
SPME Portable Field Sampler, coating PDMS
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm(PDMS, needle size 24 ga, for use with manual holder
Supelco
SPME-OC Fiber Assembly, Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 75 μm(Coating thickness includes 65 μm coating + 10 μm OC (overcoating), needle size 23 ga, pkg of 3 ea, fiber L 1 cm
Sigma-Aldrich
4-Hydroxy-2,5-dimethyl-3(2H)-furanone, ≥99.0% (GC)
Supelco
SPME fiber assortment kit 4, for use with manual holder, size 24 ga
Supelco
SPME fiber assortment kit 4, for use with autosampler, needle size 23 ga