Merck
1,2,3-Triazole derivatives as new cannabinoid CB1 receptor antagonists.
Bioorganic & medicinal chemistry letters (2008-12-20)
Duen-Ren Hou, Safiul Alam, Ting-Chun Kuan, Mani Ramanathan, Tsung-Pang Lin, Ming-Shiu Hung
ABSTRACT
This letter reports the new entry of novel 1,2,3-triazole derivatives as CB1 receptor antagonists. The design, synthesis and biological evaluation of N1 and N2 substituted 1,2,3-trizoles are described. The N2 substituted, symmetrical 1,2,3-triazoles are more potent ligands than the unsymmetrical analogues. The in vitro activity of these triazoles is further improved by inserting a methylene group between the central core and the carbonyl side chain. The most potent antagonists prepared in this series (IC(50)<20 nM) are the triazoles containing benzyl amides. These triazoles also show excellent selectivity between CB1 and CB2 receptors (IC(50)>10 microM for CB2; CB2/CB1>1000).
Sigma-Aldrich
Phenyl bromoacetate, 98%