Merck
  • Assessment of capillary anion exchange ion chromatography tandem mass spectrometry for the quantitative profiling of the phosphometabolome and organic acids in biological extracts.
Assessment of capillary anion exchange ion chromatography tandem mass spectrometry for the quantitative profiling of the phosphometabolome and organic acids in biological extracts.
Journal of chromatography. A (2014-12-03)
Hans F N Kvitvang, Kåre A Kristiansen, Per Bruheim
ABSTRACT
Metabolic profiling has become an important tool in biological research, and the chromatographic separation of metabolites coupled with mass spectrometric detection is the most frequently used approach for such studies. The establishment of robust chromatographic methods for comprehensive coverage of the anionic metabolite pool is especially challenging. In this study, the development of a capillary ion exchange chromatography (capIC) - negative ESI tandem mass spectrometry (MS/MS) workflow for the quantitative profiling of the phosphometabolome (e.g., sugar phosphates and nucleotides) is presented. The chromatographic separation and MS/MS conditions were optimized, and the precision of repetitive injections and accuracy in terms of error percentage to true concentration were assessed. The precision is excellent for a capillary flow system with an average CV% of 8.5% for a 50-fmol standard injection and in the lower 2.4-4.4% range for higher concentrations (500-7,500 fmol). The limit of detection (LOD) ranges from 1 to 100 nM (5-500 fmol injected on column), and the limit of quantitation (LOQ) ranges from 1 to 500 nM (5-2,500 fmol injected on column). A fast gradient method with the injection of 50% methanol in water between analytical samples is needed to eliminate carry-over and ensure optimal re-equilibration of the column. Finally, the quantitative applicability of the system was tested on real biological matrices using the constant-volume standard addition method (SAM). Extracts of the human kidney Hek293 cell line were spiked with increasing concentrations of standards to determine the concentration of each metabolite in the sample. Forty-four metabolites were detected with an average uncertainty of 4.1%. Thus, the capIC-MS/MS method exhibits excellent selectivity, sensitivity and precision for the quantitative profiling of the phosphometabolome.
Sigma-Aldrich
Fetal Bovine Serum, non-USA origin, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Guanosine 5′-monophosphate disodium salt hydrate, from yeast, ≥99%